Assay Details
Target Gene Details
Entrez Gene ID: | 1800 |
Gene Name: | dipeptidase 1 (renal) |
Gene Aliases: |
MBD1, MDP, RDP |
Location: |
Chr.16:89613308-89641540 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DPEP1 | NM_001128141.2 | NP_001121613.1 | ||
NM_004413.3 | NP_004404.1 | |||
XM_005256285.4 | XP_005256342.1 | |||
XM_005256286.3 | XP_005256343.1 | |||
XM_011522925.2 | XP_011521227.1 | |||
XM_017023009.1 | XP_016878498.1 | |||
AK309577.1 | ||||
BC017023.1 | AAH17023.1 | |||
BM152944.1 | ||||
BT006664.1 | AAP35310.1 | |||
CR457404.1 | CAG33685.1 | |||
D13138.1 | BAA02431.1 | |||
J05257.1 | AAB59410.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2762221 | Chr.16:89610380 - 90039318 on Build GRCh38 | Gain+Loss | GAS8-AS1 AFG3L1P ZNF276 CDK10 SPATA2L TCF25 SPATA33 SPIRE2 CENPBD1 TUBB3 VPS9D1 MC1R VPS9D1-AS1 LOC107984817 DEF8 GAS8 DPEP1 LOC105371419 FANCA CHMP1A DBNDD1 |
nsv952078 | Chr.16:89602793 - 89640192 on Build GRCh38 | Deletion | DPEP1 |
nsv428331 | Chr.16:89216635 - 90185980 on Build GRCh38 | Gain+Loss | LOC101927817 ZNF778 URAHP ZNF276 TCF25 SPIRE2 VPS9D1 PRDM7 LOC105371414 VPS9D1-AS1 FAM157C DEF8 DPEP1 LOC105371419 CHMP1A GAS8-AS1 AFG3L1P CDK10 SPATA2L ANKRD11 SPATA33 CENPBD1 TUBB3 MC1R SPG7 LOC107984817 LOC100287036 SNORD68 RPL13 GAS8 FANCA TUBB8P7 CPNE7 DBNDD1 |
nsv573739 | Chr.16:89625890 - 89696855 on Build GRCh38 | Gain | CDK10 SPATA2L SPATA33 DPEP1 CHMP1A |
nsv1063483 | Chr.16:89347250 - 89661187 on Build GRCh38 | Gain | LOC101927817 ANKRD11 SNORD68 SPATA33 RPL13 DPEP1 CHMP1A CPNE7 SPG7 |
nsv482951 | Chr.16:88633593 - 90228345 on Build GRCh38 | Loss | ZNF778 TCF25 SNAI3-AS1 LOC100129697 RNF166 PRDM7 MVD LOC105371414 VPS9D1-AS1 FAM157C LOC101927793 DPEP1 PIEZO1 LOC105371419 LOC105371409 CHMP1A AFG3L1P CDK10 SPATA33 CTU2 MC1R LOC339059 SNAI3 LOC107987238 LOC100287036 MIR4722 LOC400558 SNORD68 RPL13 GAS8 CBFA2T3 CPNE7 LOC101927817 URAHP ZNF276 SPIRE2 VPS9D1 APRT TRAPPC2L DEF8 PABPN1L CDT1 ACSF3 LOC100289580 GAS8-AS1 SPATA2L ANKRD11 CENPBD1 TUBB3 SPG7 CDH15 SLC22A31 LOC107984817 IL17C LINC00304 FANCA TUBB8P7 CYBA DBNDD1 GALNS |
nsv509639 | Chr.16:89513538 - 89775915 on Build GRCh38 | Insertion | ZNF276 CDK10 SPATA2L SPATA33 VPS9D1 SPG7 VPS9D1-AS1 SNORD68 RPL13 DPEP1 FANCA CHMP1A CPNE7 |
nsv1066382 | Chr.16:89537178 - 89661187 on Build GRCh38 | Gain | SNORD68 SPATA33 RPL13 DPEP1 CHMP1A CPNE7 SPG7 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |