Assay Details
Target Gene Details
Entrez Gene ID: | 8509 |
Gene Name: | N-deacetylase and N-sulfotransferase 2 |
Gene Aliases: |
HSST2, NST2 |
Location: |
Chr.10:73801911-73811831 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDST2 | NM_003635.3 | NP_003626.1 | ||
XM_005270255.3 | XP_005270312.1 | |||
XM_005270256.3 | XP_005270313.1 | |||
XM_011540310.2 | XP_011538612.1 | |||
XM_017016857.1 | XP_016872346.1 | |||
AB208870.1 | 2 | 1344 | BAD92107.1 | |
AK293527.1 | ||||
AK298957.1 | ||||
AK300520.1 | 1 | 94 | ||
AK301251.1 | ||||
AK303648.1 | ||||
AK310580.1 | 1 | 57 | ||
BC035711.1 | AAH35711.1 | |||
BC110588.1 | AAI10589.1 | |||
BC110589.1 | AAI10590.1 | |||
U36601.1 | AAC27120.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv46n21 | Chr.10:73790158 - 73933937 on Build GRCh38 | Loss | C10orf55 ZSWIM8-AS1 NDST2 CAMK2G ZSWIM8 PLAU |
nsv551517 | Chr.10:73799109 - 73898823 on Build GRCh38 | Loss | ZSWIM8-AS1 NDST2 CAMK2G ZSWIM8 |
nsv1035634 | Chr.10:73724401 - 73907300 on Build GRCh38 | Gain | BMS1P4 FUT11 ZSWIM8-AS1 GLUD1P3 NDST2 CAMK2G ZSWIM8 CHCHD1 SEC24C |
nsv831912 | Chr.10:73649351 - 73840445 on Build GRCh38 | Gain | BMS1P4 AGAP5 FUT11 ZSWIM8-AS1 GLUD1P3 NDST2 CAMK2G ZSWIM8 SYNPO2L CHCHD1 SEC24C |
nsv825468 | Chr.10:73715689 - 73922563 on Build GRCh38 | Gain | BMS1P4 FUT11 C10orf55 ZSWIM8-AS1 GLUD1P3 NDST2 CAMK2G ZSWIM8 CHCHD1 SEC24C PLAU |
nsv551516 | Chr.10:73749515 - 73847410 on Build GRCh38 | Loss | FUT11 ZSWIM8-AS1 NDST2 CAMK2G ZSWIM8 CHCHD1 SEC24C |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |