Assay Details
Target Gene Details
Entrez Gene ID: | 9970 |
Gene Name: | nuclear receptor subfamily 1 group I member 3 |
Gene Aliases: |
CAR, CAR1, MB67 |
Location: |
Chr.1:161229666-161238623 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NR1I3 | NM_001077469.2 | NP_001070937.1 | ||
NM_001077470.2 | NP_001070938.1 | |||
NM_001077471.2 | NP_001070939.1 | |||
NM_001077472.2 | NP_001070940.1 | |||
NM_001077473.2 | NP_001070941.1 | |||
NM_001077474.2 | NP_001070942.1 | |||
NM_001077475.2 | NP_001070943.1 | |||
NM_001077476.2 | NP_001070944.1 | |||
NM_001077477.2 | NP_001070945.1 | |||
NM_001077478.2 | NP_001070946.1 | |||
NM_001077479.2 | NP_001070947.1 | |||
NM_001077480.2 | NP_001070948.1 | |||
NM_001077481.2 | NP_001070949.1 | |||
NM_001077482.2 | NP_001070950.1 | |||
NM_005122.4 | NP_005113.1 | |||
XM_005245693.4 | 1 | 478 | XP_005245750.1 | |
XM_005245694.4 | 1 | 477 | XP_005245751.1 | |
XM_011510237.2 | 1 | 475 | XP_011508539.1 | |
AK303757.1 | ||||
AK303796.1 | 1 | 99 | ||
AK316208.1 | ||||
BC069626.1 | AAH69626.1 | |||
CB124867.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |