Assay Details
Target Gene Details
Entrez Gene ID: | 522 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial Fo complex subunit F6 |
Gene Aliases: |
ATP5, ATP5A, ATPM, CF6, F6 |
Location: |
Chr.21:25724480-25735654 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATP5J | NM_001003696.1 | NP_001003696.1 | ||
NM_001003697.1 | NP_001003697.1 | |||
NM_001003701.1 | 1 | 604 | NP_001003701.1 | |
NM_001003703.1 | 1 | 604 | NP_001003703.1 | |
NM_001320266.1 | 1 | 604 | NP_001307195.1 | |
NM_001320267.1 | 1 | 604 | NP_001307196.1 | |
NM_001685.4 | NP_001676.2 | |||
AY544129.1 | AAT11160.1 | |||
BC001178.1 | AAH01178.1 | |||
BG938438.1 | ||||
BU174560.1 | ||||
HY064673.1 |
Target Gene Details
Entrez Gene ID: | 2551 |
Gene Name: | GA binding protein transcription factor alpha subunit |
Gene Aliases: |
E4TF1-60, E4TF1A, NFT2, NRF2, NRF2A, RCH04A07 |
Location: |
Chr.21:25734947-25772460 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GABPA | NM_001197297.1 | NP_001184226.1 | ||
NM_002040.3 | NP_002031.2 | |||
BX647755.1 | ||||
CD674081.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1064561 | Chr.21:25714853 - 25750046 on Build GRCh38 | Loss | ATP5J JAM2 GABPA |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |