Assay Details
Target Gene Details
Entrez Gene ID: | 79777 |
Gene Name: | acyl-CoA binding domain containing 4 |
Gene Aliases: |
HMFT0700 |
Location: |
Chr.17:45132600-45144181 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv953903 | Chr.17:45081533 - 45148633 on Build GRCh38 | Deletion | MIR6784 HEXIM1 ACBD4 PLCD3 NMT1 |
nsv828022 | Chr.17:45137483 - 45160552 on Build GRCh38 | Gain | HEXIM2 HEXIM1 ACBD4 |
nsv527788 | Chr.17:44899824 - 45226241 on Build GRCh38 | Loss | LOC107987243 C1QL1 CCDC103 HEXIM2 FMNL1 ACBD4 GFAP PLCD3 KIF18B NMT1 MIR6783 LOC339192 MIR6784 HEXIM1 LOC105371795 DCAKD |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |