Assay Details
Target Gene Details
Entrez Gene ID: | 5653 |
Gene Name: | kallikrein related peptidase 6 |
Gene Aliases: |
Bssp, Klk7, PRSS18, PRSS9, SP59, hK6 |
Location: |
Chr.19:50958631-50969673 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLK6 | NM_001012964.2 | 6 | 1309 | NP_001012982.1 |
NM_001012965.2 | 5 | 1261 | NP_001012983.1 | |
NM_001319948.1 | 6 | 955 | NP_001306877.1 | |
NM_001319949.1 | 5 | 1152 | NP_001306878.1 | |
NM_002774.3 | 7 | 1112 | NP_002765.1 | |
AF013988.1 | 7 | 1014 | AAB66483.1 | |
AY279383.1 | 6 | 956 | ||
AY318867.1 | 6 | 1118 | AAP82446.1 | |
AY318869.1 | 6 | 1104 | AAP82448.1 | |
BC015525.2 | 6 | 1085 | AAH15525.1 | |
D78203.1 | 7 | 1023 | BAA11306.1 | |
U62801.1 | 7 | 1113 | AAB07113.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |