Assay Details
Target Gene Details
Entrez Gene ID: | 283849 |
Gene Name: | exocyst complex component 3 like 1 |
Gene Aliases: |
EXOC3L |
Location: |
Chr.16:67184379-67190204 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EXOC3L1 | NM_178516.3 | 5 | 1126 | NP_848611.2 |
AK092858.1 | 5 | 1018 | BAC03991.1 | |
AK092906.1 | 5 | 890 | BAC04000.1 | |
AK292004.1 | 5 | 1035 | ||
AK300915.1 | 5 | 911 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain | DPEP3 NRN1L CENPT KIAA0895L KCTD19 LOC100505942 ELMO3 SLC12A4 PSKH1 ATP6V0D1 THAP11 FBXL8 PARD6A EDC4 LCAT E2F4 CTCF LOC105369155 FAM65A PSMB10 C16orf86 LRRC36 LOC107984813 CARMIL2 HSD11B2 LOC100131303 SLC9A5 ENKD1 TPPP3 FHOD1 EXOC3L1 GFOD2 ZDHHC1 TMEM208 LRRC29 HSF4 ACD AGRP DDX28 TSNAXIP1 DUS2 NUTF2 RANBP10 MIR328 CTRL NOL3 DPEP2 PLEKHG4 TRADD |
nsv827707 | Chr.16:67148355 - 67289296 on Build GRCh38 | Loss | SLC9A5 E2F4 FHOD1 KIAA0895L EXOC3L1 LOC105369155 MIR328 ELMO3 C16orf70 NOL3 FBXL8 TMEM208 PLEKHG4 LRRC29 HSF4 B3GNT9 TRADD |
nsv952044 | Chr.16:67141198 - 67244397 on Build GRCh38 | Deletion | E2F4 FHOD1 KIAA0895L EXOC3L1 LOC105369155 MIR328 ELMO3 C16orf70 NOL3 FBXL8 TMEM208 LRRC29 HSF4 B3GNT9 TRADD |
nsv1160429 | Chr.16:67161549 - 67220197 on Build GRCh38 | Deletion | ELMO3 NOL3 E2F4 KIAA0895L EXOC3L1 FBXL8 LOC105369155 MIR328 LRRC29 HSF4 |
nsv457513 | Chr.16:67181417 - 67282697 on Build GRCh38 | Loss | ELMO3 SLC9A5 E2F4 FHOD1 KIAA0895L EXOC3L1 LOC105369155 TMEM208 PLEKHG4 MIR328 LRRC29 |
nsv524492 | Chr.16:67174030 - 67294550 on Build GRCh38 | Loss | SLC9A5 E2F4 FHOD1 KIAA0895L KCTD19 EXOC3L1 LOC105369155 MIR328 ELMO3 NOL3 TMEM208 PLEKHG4 LRRC29 |
nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss | DPEP3 NRN1L CENPT KIAA0895L KCTD19 LOC100505942 ELMO3 SLC12A4 PSKH1 ATP6V0D1 THAP11 FBXL8 PARD6A EDC4 LCAT E2F4 CTCF LOC105369155 C16orf70 FAM65A PSMB10 C16orf86 LRRC36 LOC107984813 CARMIL2 HSD11B2 LOC100131303 SLC9A5 ENKD1 TPPP3 FHOD1 EXOC3L1 GFOD2 ZDHHC1 TMEM208 LRRC29 HSF4 B3GNT9 ACD AGRP DDX28 TSNAXIP1 DUS2 NUTF2 RANBP10 MIR328 CTRL NOL3 DPEP2 PLEKHG4 TRADD |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |