Assay Details
Target Gene Details
Entrez Gene ID: | 1800 |
Gene Name: | dipeptidase 1 (renal) |
Gene Aliases: |
MBD1, MDP, RDP |
Location: |
Chr.16:89613308-89641540 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DPEP1 | NM_001128141.2 | 4 | 501 | NP_001121613.1 |
NM_004413.3 | 4 | 589 | NP_004404.1 | |
XM_005256285.4 | 4 | 539 | XP_005256342.1 | |
XM_005256286.3 | 4 | 451 | XP_005256343.1 | |
XM_011522925.2 | 4 | 549 | XP_011521227.1 | |
XM_017023009.1 | 4 | 612 | XP_016878498.1 | |
AK309577.1 | 3 | 365 | ||
BC017023.1 | 3 | 420 | AAH17023.1 | |
BM152944.1 | ||||
BT006664.1 | 3 | 298 | AAP35310.1 | |
CR457404.1 | 3 | 298 | CAG33685.1 | |
D13138.1 | 3 | 341 | BAA02431.1 | |
J05257.1 | 4 | 593 | AAB59410.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2762221 | Chr.16:89610380 - 90039318 on Build GRCh38 | Gain+Loss | DEF8 FANCA VPS9D1 DPEP1 TCF25 VPS9D1-AS1 CHMP1A AFG3L1P LOC107984817 SPATA33 CENPBD1 GAS8-AS1 TUBB3 MC1R LOC105371419 CDK10 GAS8 SPIRE2 SPATA2L ZNF276 DBNDD1 |
nsv952078 | Chr.16:89602793 - 89640192 on Build GRCh38 | Deletion | DPEP1 |
nsv1930 | Chr.16:89633876 - 89640750 on Build GRCh38 | Insertion | DPEP1 |
nsv428331 | Chr.16:89216635 - 90185980 on Build GRCh38 | Gain+Loss | CPNE7 SPG7 LOC105371414 VPS9D1 FAM157C LOC100287036 AFG3L1P CENPBD1 URAHP ANKRD11 TUBB3 MC1R CDK10 RPL13 SNORD68 SPATA2L DEF8 PRDM7 LOC101927817 FANCA DPEP1 TCF25 VPS9D1-AS1 CHMP1A LOC107984817 SPATA33 GAS8-AS1 LOC105371419 GAS8 TUBB8P7 SPIRE2 ZNF778 ZNF276 DBNDD1 |
nsv573739 | Chr.16:89625890 - 89696855 on Build GRCh38 | Gain | CDK10 DPEP1 SPATA2L CHMP1A SPATA33 |
nsv1063483 | Chr.16:89347250 - 89661187 on Build GRCh38 | Gain | ANKRD11 LOC101927817 CPNE7 SPG7 RPL13 DPEP1 SNORD68 CHMP1A SPATA33 |
nsv482951 | Chr.16:88633593 - 90228345 on Build GRCh38 | Loss | LOC100129697 SPG7 LOC105371414 TRAPPC2L CBFA2T3 CENPBD1 SLC22A31 ANKRD11 MC1R CDK10 RPL13 PIEZO1 CYBA SPATA2L CDH15 DEF8 PRDM7 LOC101927817 TCF25 VPS9D1-AS1 LOC107984817 SPATA33 GAS8-AS1 LOC105371419 GAS8 LOC100289580 RNF166 SNAI3-AS1 ZNF276 IL17C CPNE7 LOC105371409 SNAI3 VPS9D1 FAM157C LOC100287036 LOC400558 AFG3L1P MVD APRT URAHP TUBB3 SNORD68 LINC00304 FANCA ACSF3 DPEP1 CDT1 LOC339059 PABPN1L CHMP1A CTU2 LOC107987238 MIR4722 LOC101927793 TUBB8P7 GALNS SPIRE2 ZNF778 DBNDD1 |
nsv509639 | Chr.16:89513538 - 89775915 on Build GRCh38 | Insertion | CPNE7 SPG7 FANCA VPS9D1 DPEP1 VPS9D1-AS1 CHMP1A SPATA33 CDK10 RPL13 SNORD68 SPATA2L ZNF276 |
nsv1066382 | Chr.16:89537178 - 89661187 on Build GRCh38 | Gain | CPNE7 SPG7 RPL13 DPEP1 SNORD68 CHMP1A SPATA33 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |