Assay Details
Target Gene Details
Entrez Gene ID: | 79042 |
Gene Name: | tRNA splicing endonuclease subunit 34 |
Gene Aliases: |
LENG5, PCH2C, SEN34, SEN34L |
Location: |
Chr.19:54189517-54194536 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TSEN34 | NM_001282333.1 | NP_001269262.1 | ||
NM_024075.4 | 1 | 85 | NP_076980.2 | |
XM_005278290.4 | XP_005278347.1 | |||
XM_011527294.2 | XP_011525596.1 | |||
XM_011527295.1 | XP_011525597.1 | |||
AK025929.1 | BAB15284.1 | |||
AK054944.1 | ||||
AK225385.1 | ||||
AK226013.1 | ||||
AK300054.1 | ||||
DR763016.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1066425 | Chr.19:54182441 - 54237431 on Build GRCh38 | Gain | MBOAT7 TSEN34 LILRB3 RPS9 LILRA6 |
nsv9751 | Chr.19:54158885 - 54193528 on Build GRCh38 | Gain | MBOAT7 TSEN34 TMC4 LENG1 |
nsv2546 | Chr.19:54182614 - 54270671 on Build GRCh38 | Deletion | MBOAT7 TSEN34 LILRB3 RPS9 LILRA6 LILRB5 |
nsv953613 | Chr.19:54097214 - 54198733 on Build GRCh38 | Deletion | NDUFA3 PRPF31 MBOAT7 TSEN34 CNOT3 TMC4 TFPT OSCAR LENG1 |
esv2718812 | Chr.19:53184636 - 54322450 on Build GRCh38 | Deletion | MIR518B MIR373 VN1R4 MIR519E MIR516B2 MIR523 TSEN34 MIR1323 MIR527 MIR520H ZNF765 RPS9 MIR935 MIR372 MIR519A1 PRKCG ZNF677 ZNF761 ZNF845 PRPF31 MIR1283-2 ZNF525 TFPT MIR512-2 LILRB5 MIR520G MIR520F MIR521-2 MIR371B LILRA5 MIR518E MIR526A1 MBOAT7 MIR519D LOC105372457 LILRA6 MIR515-2 MIR521-1 DPRX MIR526A2 MIR525 MIR526B MIR371A TMC4 ZNF331 MIR517B MIR518F CACNG6 LILRB2 MIR520A NDUFA3 MIR518C VSTM1 MIR517A MIR520B MIR498 MIR519B BIRC8 MYADM MIR516A1 LENG1 MIR517C MIR519C TARM1 MIR1283-1 MIR524 VN1R2 CNOT3 MIR512-1 OSCAR NLRP12 MIR522 MIR518A1 MIR516A2 MIR516B1 FAM90A27P MIR520C ZNF813 MIR4752 MIR520E CACNG8 MIR520D MIR518A2 CACNG7 TPM3P9 LILRB3 LOC284379 MIR519A2 MIR518D ZNF665 MIR515-1 ZNF818P |
nsv510774 | Chr.19:54158011 - 54269511 on Build GRCh38 | Deletion | MBOAT7 TSEN34 LILRB3 TMC4 RPS9 LILRA6 LENG1 LILRB5 |
esv2758771 | Chr.19:54163847 - 54954956 on Build GRCh38 | Gain+Loss | LENG8-AS1 CDC42EP5 TSEN34 KIR3DL1 KIR2DL1 RPS9 LILRA2 KIR3DL3 FCAR LOC101928804 LOC102724300 NLRP7 LILRP2 LILRB5 KIR2DL3 LENG9 KIR2DL4 LILRA1 LILRB4 KIR2DS4 LILRA5 MBOAT7 LAIR1 LILRA6 MIR8061 MIR4752 KIR3DL2 LENG8 KIR3DX1 LILRA4 LILRB3 TMC4 NCR1 LAIR2 TTYH1 LILRB1 LILRB2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |