Assay Details
Target Gene Details
Entrez Gene ID: | 5005 |
Gene Name: | orosomucoid 2 |
Gene Aliases: |
AGP-B, AGP-B', AGP2 |
Location: |
Chr.9:114329789-114333256 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ORM2 | NM_000608.2 | NP_000599.1 | ||
AK303760.1 | ||||
AK312226.1 | ||||
BC015964.1 | AAH15964.1 | |||
BC056239.1 | ||||
CR456930.1 | CAG33211.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv2181e212 | Chr.9:114317539 - 114335437 on Build GRCh38 | Gain | AKNA ORM1 ORM2 |
nsv968749 | Chr.9:114324831 - 114335180 on Build GRCh38 | Duplication | AKNA ORM1 ORM2 |
esv3576701 | Chr.9:114320518 - 114331888 on Build GRCh38 | Gain | ORM1 ORM2 |
esv3576696 | Chr.9:114324735 - 114335437 on Build GRCh38 | Gain | AKNA ORM1 ORM2 |
dgv33e196 | Chr.9:114030664 - 114604363 on Build GRCh38 | Duplication | WHRN COL27A1 MIR455 AKNA ORM1 AMBP ZNF618 ORM2 KIF12 ATP6V1G1 TMEM268 |
nsv1110578 | Chr.9:114322720 - 114334420 on Build GRCh38 | Duplication | AKNA ORM1 ORM2 |
dgv949n27 | Chr.9:114283521 - 114343856 on Build GRCh38 | Gain | COL27A1 AKNA ORM1 ORM2 |
esv2462819 | Chr.9:114327942 - 114335805 on Build GRCh38 | Deletion | AKNA ORM2 |
nsv615247 | Chr.9:114330749 - 114331675 on Build GRCh38 | Loss | ORM2 |
esv22321 | Chr.9:114320556 - 114336003 on Build GRCh38 | Gain | AKNA ORM1 ORM2 |
dgv948n27 | Chr.9:114193988 - 114693895 on Build GRCh38 | Gain | WHRN LOC100505478 COL27A1 MIR455 AKNA ORM1 ORM2 ATP6V1G1 TMEM268 |
esv2664309 | Chr.9:114325228 - 114332005 on Build GRCh38 | Deletion | ORM1 ORM2 |
esv2752277 | Chr.9:114323991 - 114338965 on Build GRCh38 | Gain | AKNA ORM1 ORM2 |
esv2761285 | Chr.9:114049617 - 114340988 on Build GRCh38 | Gain | COL27A1 MIR455 AKNA ORM1 AMBP ZNF618 ORM2 KIF12 |
nsv509319 | Chr.9:114297186 - 114380605 on Build GRCh38 | Insertion | COL27A1 AKNA ORM1 ORM2 |
dgv7706n100 | Chr.9:114182745 - 114697870 on Build GRCh38 | Gain | WHRN LOC100505478 COL27A1 MIR455 AKNA ORM1 ORM2 ATP6V1G1 TMEM268 |
esv3621554 | Chr.9:114316410 - 114335883 on Build GRCh38 | Loss | AKNA ORM1 ORM2 |
nsv471320 | Chr.9:114284841 - 114346385 on Build GRCh38 | Gain | COL27A1 AKNA ORM1 ORM2 |
nsv615233 | Chr.9:114167820 - 114486199 on Build GRCh38 | Gain | WHRN COL27A1 MIR455 AKNA ORM1 ORM2 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |