Assay Details
Target Gene Details
Entrez Gene ID: | 6446 |
Gene Name: | serum/glucocorticoid regulated kinase 1 |
Gene Aliases: |
SGK |
Location: |
Chr.6:134169246-134318112 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SGK1 | NM_001143676.1 | 14 | 2303 | NP_001137148.1 |
NM_001143677.1 | 12 | 1554 | NP_001137149.1 | |
NM_001143678.1 | 12 | 1733 | NP_001137150.1 | |
NM_001291995.1 | 11 | 1377 | NP_001278924.1 | |
NM_005627.3 | 12 | 1509 | NP_005618.2 | |
XM_011536071.1 | 15 | 2085 | XP_011534373.1 | |
AF153609.1 | 12 | 1462 | AAD41091.1 | |
AK055077.1 | 14 | 2303 | ||
AK098509.1 | 12 | 1477 | ||
AK293383.1 | 12 | 1543 | ||
AK295422.1 | 11 | 1345 | ||
AK298688.1 | 12 | 1792 | ||
BC001263.1 | 12 | 1462 | AAH01263.1 | |
BX649005.1 | 2 | 4579 | CAH10381.1 | |
FM205707.1 | 12 | 1497 | ||
FM205708.1 | 12 | 1532 | ||
FM205709.1 | 14 | 2152 | ||
FM205710.1 | 11 | 1621 | ||
Y10032.1 | 12 | 1462 | CAA71138.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |