Assay Details
Target Gene Details
Entrez Gene ID: | 282809 |
Gene Name: | POC1 centriolar protein B |
Gene Aliases: |
CORD20, PIX1, TUWD12, WDR51B |
Location: |
Chr.12:89419718-89526262 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POC1B | NM_001199777.1 | NP_001186706.1 | ||
NM_172240.2 | NP_758440.1 | |||
NR_037659.1 | ||||
NR_037660.1 | ||||
AB053266.1 | BAB69057.1 | |||
AK027829.1 | ||||
AK074772.1 | BAC11198.1 | |||
AK123106.1 | ||||
AK123625.1 | BAC85666.1 | |||
AK295596.1 | ||||
AL832918.1 | CAH10775.1 | |||
BC026080.2 | AAH26080.1 | |||
BX648293.1 | ||||
EF011619.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |