Assay Details
Target Gene Details
Entrez Gene ID: | 64223 |
Gene Name: | MTOR associated protein, LST8 homolog |
Gene Aliases: |
GBL, GbetaL, LST8, POP3, WAT1 |
Location: |
Chr.16:2205177-2209417 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MLST8 | NM_001199173.1 | 9 | 1558 | NP_001186102.1 |
NM_001199174.1 | 9 | 1397 | NP_001186103.1 | |
NM_001199175.1 | 9 | 1585 | NP_001186104.1 | |
NM_022372.4 | 9 | 1588 | NP_071767.3 | |
XM_005255475.3 | 9 | 1364 | XP_005255532.2 | |
XM_005255478.2 | 9 | 1569 | XP_005255535.1 | |
XM_005255479.2 | 9 | 1496 | XP_005255536.1 | |
XM_017023548.1 | 9 | 1553 | XP_016879037.1 | |
XM_017023549.1 | 9 | 1294 | XP_016879038.1 | |
XM_017023551.1 | 9 | 1603 | XP_016879040.1 | |
AF195883.1 | 6 | 909 | AAF04308.1 | |
AK021536.1 | 9 | 1557 | ||
AK022227.1 | 8 | 1242 | BAB13990.1 | |
AK098762.1 | 9 | 1493 | ||
AK123195.1 | 2 | 1363 | ||
AK302201.1 | ||||
AY223837.1 | 8 | 1398 | AAO73410.1 | |
BC001313.2 | 9 | 1278 | AAH01313.1 | |
BC017119.1 | 8 | 1289 | AAH17119.1 | |
BC052292.1 | 8 | 1812 | AAH52292.1 | |
BC088354.1 | 9 | 1306 | AAH88354.1 | |
BQ014640.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | RPS2 LOC101929280 SNORA78 DECR2 IFT140 SYNGR3 NTHL1 MRPS34 CHTF18 RPL3L TBL3 E4F1 NHLRC4 MIR6511B1 WASIR2 MIR662 HN1L TPSB2 NUBP2 SNRNP25 LINC00254 PDIA2 MIR3677 NME3 LOC652276 LMF1 LOC105371038 CRAMP1 CEMP1 LOC105371045 CAPN15 NOXO1 LOC105371047 SNORD60 SSTR5 ABCA17P NPW MEIOB NPRL3 CCNF RGS11 CCDC78 CCDC154 METTL26 UNKL TBC1D24 ZNF598 SOX8 PRR25 WFIKKN1 NARFL SSTR5-AS1 RPUSD1 TPSAB1 ERVK13-1 HBA1 ABCA3 NTN3 RAB11FIP3 SNORA10 MIR1225 MIR5587 ECI1 ATP6V0C NDUFB10 RHBDL1 MIR6768 MRPL28 GFER DDX11L10 LOC105371049 WDR90 LOC100287175 TPSD1 LOC105371184 HBZ PIGQ GNPTG IGFALS MIR4516 MIR940 BRICD5 DNASE1L2 LOC105371046 MIR3178 HS3ST6 METRN CASKIN1 MIR3176 FAM173A SLC9A3R2 TSR3 TMEM204 TPSG1 MIR6767 SNHG9 UBE2I EME2 RPL23AP5 MSRB1 ARHGDIG PRSS29P LOC106660606 MLST8 CACNA1H POLR3K HBQ1 LOC107987233 LINC00235 C16orf91 MIR3180-5 BAIAP3 STUB1 HBA2 MIR4717 TSC2 KCTD5 HAGHL FAM234A LMF1-AS1 PGP MIR6859-4 LOC100134368 RNF151 MPG GNG13 RHBDF1 TMEM8A PKD1 WDR24 LUC7L RAB40C NME4 FLJ42627 C1QTNF8 TELO2 HBM MIR3177 RAB26 JMJD8 AXIN1 SPSB3 PDPK1 SNHG19 HAGH RHOT2 C16orf59 RNPS1 SNORA64 TRAF7 CLCN7 PTX4 MAPK8IP3 LOC107984876 LOC729652 AMDHD2 FAHD1 MCRIP2 PRSS27 PRR35 FBXL16 MSLN |
nsv471072 | Chr.16:1971892 - 2317508 on Build GRCh38 | Loss | CASKIN1 SLC9A3R2 PKD1 ZNF598 SYNGR3 NTHL1 TBL3 ABCA3 RAB26 E4F1 MIR6511B1 MIR1225 SNHG19 ECI1 RNPS1 LOC106660606 MLST8 TRAF7 MIR3677 GFER MIR3180-5 MIR4717 TSC2 LOC105371049 NOXO1 SNORD60 MIR4516 MIR940 BRICD5 PGP NPW DNASE1L2 |
dgv4861n54 | Chr.16:2083900 - 2246850 on Build GRCh38 | Loss | MIR6511B1 MIR1225 CASKIN1 SNHG19 ECI1 MLST8 TRAF7 PKD1 MIR3180-5 TSC2 LOC105371049 SNORD60 MIR4516 BRICD5 RAB26 PGP E4F1 DNASE1L2 |
dgv4863n54 | Chr.16:2141733 - 2274302 on Build GRCh38 | Loss | CASKIN1 SNHG19 ECI1 RNPS1 LOC106660606 MLST8 TRAF7 MIR3677 SNORD60 MIR940 BRICD5 RAB26 PGP E4F1 DNASE1L2 |
nsv1711 | Chr.16:2199093 - 2232650 on Build GRCh38 | Insertion | MLST8 BRICD5 PGP E4F1 |
nsv457351 | Chr.16:2156590 - 2274302 on Build GRCh38 | Loss | CASKIN1 ECI1 RNPS1 LOC106660606 MLST8 TRAF7 MIR3677 MIR940 BRICD5 PGP E4F1 DNASE1L2 |
nsv1052260 | Chr.16:2074546 - 2629655 on Build GRCh38 | Gain | MIR3178 CASKIN1 PKD1 TBC1D24 MIR6767 ABCA3 NTN3 RAB26 E4F1 MIR6511B1 MIR1225 PDPK1 SNHG19 ECI1 ATP6V0C C16orf59 RNPS1 LOC106660606 MLST8 TRAF7 MIR6768 MIR3677 MIR3180-5 LOC652276 MIR4717 TSC2 LOC105371049 CEMP1 LOC729652 AMDHD2 SNORD60 MIR4516 MIR940 BRICD5 PGP ABCA17P DNASE1L2 CCNF |
nsv952902 | Chr.16:2143400 - 2339999 on Build GRCh38 | Deletion | CASKIN1 SNHG19 ECI1 RNPS1 LOC106660606 MLST8 TRAF7 MIR3677 MIR4717 SNORD60 ABCA3 MIR940 BRICD5 RAB26 PGP E4F1 DNASE1L2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |