Assay Details
Target Gene Details
Entrez Gene ID: | 54496 |
Gene Name: | protein arginine methyltransferase 7 |
Gene Aliases: |
- |
Location: |
Chr.16:68310974-68358832 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRMT7 | XM_011523112.2 | 19 | 3235 | XP_011521414.1 |
XM_011523113.2 | 20 | 3362 | XP_011521415.1 | |
XM_011523115.2 | 19 | 3179 | XP_011521417.1 | |
XM_011523116.2 | 19 | 3232 | XP_011521418.1 | |
XM_011523121.2 | 17 | 3000 | XP_011521423.1 | |
XM_011523124.2 | 19 | 3288 | XP_011521426.1 | |
XM_011523125.2 | 18 | 3130 | XP_011521427.1 | |
XM_011523126.2 | 18 | 3134 | XP_011521428.1 | |
XM_011523128.2 | 19 | 3179 | XP_011521430.1 | |
XM_017023291.1 | 20 | 3299 | XP_016878780.1 | |
XM_017023292.1 | 19 | 3176 | XP_016878781.1 | |
XM_017023296.1 | 20 | 3385 | XP_016878785.1 | |
XM_017023298.1 | 18 | 3230 | XP_016878787.1 | |
XM_017023300.1 | 17 | 3140 | XP_016878789.1 | |
XM_017023301.1 | 17 | 3137 | XP_016878790.1 | |
XM_017023302.1 | 18 | 3127 | XP_016878791.1 | |
XM_017023303.1 | 19 | 3285 | XP_016878792.1 | |
XM_017023311.1 | 18 | 3083 | XP_016878800.1 |
Target Gene Details
Entrez Gene ID: | 55512 |
Gene Name: | sphingomyelin phosphodiesterase 3 |
Gene Aliases: |
NSMASE2 |
Location: |
Chr.16:68358327-68448703 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SMPD3 | NM_018667.3 | 9 | 4837 | NP_061137.1 |
XM_005256031.3 | 9 | 4930 | XP_005256088.1 | |
XM_005256032.3 | 9 | 4949 | XP_005256089.1 | |
XM_011523207.1 | 10 | 4930 | XP_011521509.1 | |
XM_011523208.2 | 8 | 5031 | XP_011521510.1 | |
XM_011523209.2 | 8 | 4814 | XP_011521511.1 | |
XM_017023405.1 | 9 | 4756 | XP_016878894.1 | |
XM_017023406.1 | 10 | 5237 | XP_016878895.1 | |
XM_017023407.1 | 10 | 4959 | XP_016878896.1 | |
XM_017023408.1 | 10 | 4913 | XP_016878897.1 | |
AK026246.1 | 1 | 995 | BAB15409.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3582280 | Chr.16:68348088 - 68360930 on Build GRCh38 | Loss | SMPD3 PRMT7 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |