Assay Details
Target Gene Details
Entrez Gene ID: | 59307 |
Gene Name: | single Ig and TIR domain containing |
Gene Aliases: |
IL-1R8, TIR8 |
Location: |
Chr.11:405716-417454 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SIGIRR | NM_001135053.1 | NP_001128525.1 | ||
NM_001135054.1 | NP_001128526.1 | |||
NM_021805.2 | NP_068577.2 | |||
XM_005253044.2 | XP_005253101.1 | |||
XM_005253045.1 | XP_005253102.1 | |||
XM_005253046.1 | XP_005253103.1 | |||
XM_005253048.4 | XP_005253105.1 | |||
XM_017018099.1 | XP_016873588.1 | |||
XM_017018100.1 | XP_016873589.1 | |||
AK025099.1 | BAB15066.1 | |||
AK093427.1 | ||||
AK172830.1 | BAD18795.1 | |||
AY358342.1 | AAQ88708.1 | |||
BC003591.1 | AAH03591.1 | |||
BC025953.1 | AAH25953.1 | |||
BC106007.1 | AAI06008.1 | |||
CR457338.1 | CAG33619.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv527327 | Chr.11:295343 - 620599 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv469923 | Chr.11:378188 - 1302334 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv552776 | Chr.11:366710 - 432436 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
nsv951268 | Chr.11:368601 - 503700 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv552770 | Chr.11:356090 - 444628 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
dgv1505n54 | Chr.11:198510 - 470331 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1159790 | Chr.11:340219 - 491334 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() |
dgv182e199 | Chr.11:349830 - 943372 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv552822 | Chr.11:403545 - 408352 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)