Assay Details
Target Gene Details
Entrez Gene ID: | 90627 |
Gene Name: | StAR related lipid transfer domain containing 13 |
Gene Aliases: |
ARHGAP37, DLC2, GT650, LINC00464 |
Location: |
Chr.13:33103135-33676835 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STARD13 | NM_001243476.2 | 18 | 5651 | NP_001230405.1 |
NM_052851.2 | 14 | 5241 | NP_443083.1 | |
NM_178006.3 | 14 | 5304 | NP_821074.1 | |
NM_178007.2 | 14 | 5211 | NP_821075.1 | |
XM_011535299.2 | 17 | 5397 | XP_011533601.1 | |
XM_017020834.1 | 14 | 5409 | XP_016876323.1 | |
XM_017020835.1 | 16 | 5330 | XP_016876324.1 | |
AL049801.1 | 14 | 5197 | CAB42562.1 | |
AY082589.1 | 14 | 5299 | AAL91648.1 | |
AY082590.1 | 14 | 5223 | AAL91649.1 | |
AY082591.1 | 14 | 5197 | AAL91650.1 | |
BX641149.1 | 4 | 2324 | CAE46065.1 | |
BX647695.1 | 18 | 5611 | CAI46026.1 | |
CB163673.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |