Assay Details
Target Gene Details
Entrez Gene ID: | 6285 |
Gene Name: | S100 calcium binding protein B |
Gene Aliases: |
NEF, S100, S100-B, S100beta |
Location: |
Chr.21:46598618-46605242 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
S100B | NM_006272.2 | 3 | 317 | NP_006263.1 |
XM_017028424.1 | 3 | 444 | XP_016883913.1 | |
BC001766.1 | 3 | 268 | AAH01766.1 | |
CR542123.1 | 2 | 206 | CAG46920.1 | |
EB386131.1 | 3 | 266 | ||
KU178319.1 | 2 | 206 | ||
KU178320.1 | 3 | 300 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv4442n100 | Chr.21:46551020 - 46677033 on Build GRCh38 | Gain | S100B DIP2A PRMT2 |
nsv587999 | Chr.21:46522003 - 46683939 on Build GRCh38 | Gain | S100B DIP2A PRMT2 |
nsv528495 | Chr.21:46595956 - 46632926 on Build GRCh38 | Loss | S100B |
nsv588002 | Chr.21:46575530 - 46653004 on Build GRCh38 | Gain | S100B PRMT2 |
esv34393 | Chr.21:46595759 - 46657900 on Build GRCh38 | Gain | S100B PRMT2 |
esv3647171 | Chr.21:46586178 - 46674643 on Build GRCh38 | Gain | S100B PRMT2 |
dgv4443n100 | Chr.21:46561965 - 46646565 on Build GRCh38 | Gain | S100B DIP2A PRMT2 |
dgv4441n100 | Chr.21:46474792 - 46677033 on Build GRCh38 | Gain | S100B DIP2A PRMT2 |
esv2758822 | Chr.21:46566757 - 46657900 on Build GRCh38 | Gain | S100B DIP2A PRMT2 |
nsv528947 | Chr.21:46566170 - 46630476 on Build GRCh38 | Gain | S100B DIP2A |
esv3647172 | Chr.21:46592879 - 46650385 on Build GRCh38 | Loss | S100B PRMT2 |
esv2760700 | Chr.21:46554142 - 46634864 on Build GRCh38 | Gain | S100B DIP2A |
nsv470910 | Chr.21:45933861 - 46665076 on Build GRCh38 | Gain | PCBP3 MCM3AP-AS1 S100B MCM3AP C21orf58 DIP2A COL6A1 COL6A2 SPATC1L LSS PRMT2 FTCD PCNT LOC101928796 DIP2A-IT1 YBEY |
nsv1065227 | Chr.21:46565742 - 46616312 on Build GRCh38 | Gain | S100B DIP2A |
nsv1057566 | Chr.21:46495670 - 46609786 on Build GRCh38 | Loss | S100B DIP2A |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |