Assay Details
Target Gene Details
Entrez Gene ID: | 7050 |
Gene Name: | TGFB induced factor homeobox 1 |
Gene Aliases: |
HPE4, TGIF |
Location: |
Chr.18:3411927-3458412 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TGIF1 | NM_001278682.1 | NP_001265611.1 | ||
NM_001278684.1 | 2 | 496 | NP_001265613.1 | |
NM_001278686.1 | NP_001265615.1 | |||
NM_003244.3 | 1 | 229 | NP_003235.1 | |
NM_173207.2 | NP_775299.1 | |||
NM_173208.2 | 2 | 499 | NP_775300.1 | |
NM_173209.2 | NP_775301.1 | |||
NM_174886.2 | NP_777480.1 | |||
XM_017025958.1 | 2 | 560 | XP_016881447.1 | |
XM_017025959.1 | XP_016881448.1 | |||
AI813434.1 | 2 | 117 | ||
AK291112.1 | ||||
AK309471.1 | 1 | 224 | ||
AU310737.1 | 1 | 229 | ||
BC000814.1 | AAH00814.1 | |||
BE296707.1 | ||||
BE906361.1 | ||||
BF446044.1 | 2 | 111 | ||
BM849453.1 | ||||
BQ437864.1 | 1 | 227 | ||
BU657748.1 | ||||
CB161219.1 | ||||
CB996365.1 | ||||
DR007147.1 | ||||
HY041619.1 | ||||
X89750.1 | 1 | 222 | CAA61897.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv527676 | Chr.18:3445828 - 3452980 on Build GRCh38 | Loss | TGIF1 |
nsv516490 | Chr.18:3448764 - 3451764 on Build GRCh38 | Loss | TGIF1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |