Assay Details
Target Gene Details
Entrez Gene ID: | 285203 |
Gene Name: | EGF domain specific O-linked N-acetylglucosamine transferase |
Gene Aliases: |
AER61, AOS4, C3orf64, EOGT1 |
Location: |
Chr.3:68975212-69013961 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EOGT | NM_001278689.1 | 18 | 2190 | NP_001265618.1 |
NM_173654.2 | 15 | 1938 | NP_775925.1 | |
NR_103826.1 | 16 | 1979 | ||
XM_005264743.2 | 18 | 2404 | XP_005264800.1 | |
XM_011533599.1 | 19 | 2458 | XP_011531901.1 | |
XM_011533600.1 | 19 | 2311 | XP_011531902.1 | |
XM_011533602.1 | 19 | 2223 | XP_011531904.1 | |
XM_011533603.1 | 19 | 2455 | XP_011531905.1 | |
XM_017006204.1 | 19 | 2308 | XP_016861693.1 | |
XM_017006205.1 | 18 | 2169 | XP_016861694.1 | |
XM_017006206.1 | 18 | 2065 | XP_016861695.1 | |
XM_017006207.1 | 11 | 904 | XP_016861696.1 | |
XM_017006209.1 | 9 | 777 | XP_016861698.1 | |
AJ868234.1 | 15 | 1514 | CAI30569.1 | |
AK091089.1 | 9 | 1168 | ||
AK123213.1 | 6 | 502 | ||
AK126187.1 | BAC86479.1 | |||
AK290356.1 | 14 | 1317 | ||
AK294101.1 | 10 | 830 | ||
AK304102.1 | 16 | 1735 | ||
AL833467.1 | 4 | 344 | ||
BC060887.1 | 15 | 1667 | AAH60887.1 | |
BX640821.1 | 14 | 1311 | CAE45897.2 | |
BX648818.1 | 18 | 1965 | ||
KC347596.1 | 15 | 1514 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |