Assay Details
Target Gene Details
Entrez Gene ID: | 64852 |
Gene Name: | terminal uridylyl transferase 1, U6 snRNA-specific |
Gene Aliases: |
PAPD2, RBM21, STARPAP, TUTase, URLC6 |
Location: |
Chr.11:62575045-62591637 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TUT1 | NM_022830.2 | 9 | 2162 | NP_073741.2 |
AB101208.1 | 9 | 2076 | ||
AK025920.1 | 5 | 1125 | BAB15282.1 | |
AK026000.1 | 9 | 2069 | BAB15314.1 | |
AK292610.1 | 9 | 2161 | ||
BC005013.1 | 5 | 1124 | AAH05013.2 | |
BC110910.1 | 9 | 2051 | AAI10911.1 | |
BC128263.1 | 9 | 2055 | ||
KU178756.1 | 5 | 993 | ||
KU178757.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3584599 | Chr.11:62495270 - 62630578 on Build GRCh38 | Gain | MTA2 ROM1 EEF1G MIR6747 GANAB B3GAT3 TUT1 EML3 AHNAK |
nsv832180 | Chr.11:62434133 - 62597356 on Build GRCh38 | Loss | MTA2 EEF1G MIR6747 TUT1 AHNAK |
nsv825939 | Chr.11:62558611 - 62643105 on Build GRCh38 | Gain | MTA2 ROM1 EEF1G MIR6747 GANAB B3GAT3 TUT1 EML3 |
nsv832182 | Chr.11:62528103 - 62695376 on Build GRCh38 | Gain+Loss | ROM1 EEF1G LBHD1 MIR6747 HNRNPUL2-BSCL2 GANAB INTS5 TUT1 C11orf98 METTL12 UQCC3 MTA2 LRRN4CL BSCL2 B3GAT3 SNORA57 UBXN1 EML3 AHNAK |
nsv555174 | Chr.11:62566624 - 62729047 on Build GRCh38 | Gain | TTC9C ROM1 EEF1G LBHD1 MIR6747 HNRNPUL2-BSCL2 GANAB INTS5 TUT1 C11orf98 METTL12 UQCC3 MTA2 HNRNPUL2 LRRN4CL BSCL2 B3GAT3 SNORA57 UBXN1 EML3 GNG3 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |