Assay Details
Target Gene Details
Entrez Gene ID: | 8624 |
Gene Name: | proteasome assembly chaperone 1 |
Gene Aliases: |
C21LRP, DSCR2, LRPC21, PAC-1, PAC1 |
Location: |
Chr.21:39175446-39183514 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PSMG1 | NM_001261824.1 | 3 | 448 | NP_001248753.1 |
NM_001320795.1 | 3 | 636 | NP_001307724.1 | |
NM_003720.3 | 3 | 448 | NP_003711.1 | |
NM_203433.2 | 3 | 448 | NP_982257.1 | |
NR_049728.1 | 2 | 341 | ||
XM_017028488.1 | 3 | 362 | XP_016883977.1 | |
AB451468.1 | 3 | 319 | ||
AF417108.1 | 2 | 212 | ||
AJ006291.1 | 3 | 382 | CAA06957.1 | |
AK001455.1 | 3 | 410 | ||
AK308614.1 | 2 | 319 | ||
AK315406.1 | 3 | 350 | ||
AY463963.1 | 3 | 319 | AAR25628.1 | |
BC003619.2 | 3 | 365 | AAH03619.1 | |
BC010424.1 | 3 | 357 | AAH10424.1 | |
BC011755.2 | 3 | 365 | AAH11755.1 | |
BC012809.2 | 3 | 365 | AAH12809.1 | |
BE779375.1 | 3 | 445 | ||
BM013494.1 | 3 | 316 | ||
BM460414.1 | 3 | 341 | ||
BM462074.1 | 3 | 380 | ||
BQ008489.1 | 5 | 697 | ||
BU543452.1 | 3 | 399 | ||
CB160221.1 | 3 | 310 | ||
CR541821.1 | 3 | 319 | CAG46620.1 | |
CR541852.1 | 3 | 319 | CAG46650.1 | |
FN152980.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |