Assay Details
Target Gene Details
Entrez Gene ID: | 79612 |
Gene Name: | N(alpha)-acetyltransferase 16, NatA auxiliary subunit |
Gene Aliases: |
NARG1L |
Location: |
Chr.13:41311205-41377031 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NAA16 | NM_001110798.1 | NP_001104268.1 | ||
NM_018527.3 | NP_060997.2 | |||
NM_024561.4 | NP_078837.3 | |||
XM_005266523.2 | XP_005266580.1 | |||
XM_006719866.2 | XP_006719929.1 | |||
XM_011535227.2 | XP_011533529.1 | |||
XM_011535228.1 | XP_011533530.1 | |||
XM_017020744.1 | XP_016876233.1 | |||
XM_017020745.1 | XP_016876234.1 | |||
XM_017020746.1 | XP_016876235.1 | |||
AK097224.1 | ||||
AK291248.1 | ||||
AL833341.1 | ||||
BC032318.1 | AAH32318.1 | |||
BC064592.1 | AAH64592.1 | |||
BC101719.1 | ||||
BC101721.1 | ||||
BE501152.1 | ||||
BX640665.1 | CAE45801.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |