Assay Details
Target Gene Details
Entrez Gene ID: | 2208 |
Gene Name: | Fc fragment of IgE receptor II |
Gene Aliases: |
BLAST-2, CD23, CD23A, CLEC4J, FCE2, IGEBF |
Location: |
Chr.19:7688757-7702755 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FCER2 | NM_001220500.1 | NP_001207429.1 | ||
NM_002002.4 | NP_001993.2 | |||
XM_005272462.4 | 1 | 659 | XP_005272519.1 | |
AK223207.1 | BAD96927.1 | |||
AK310071.1 | ||||
AK310707.1 | ||||
BC014108.2 | AAH14108.1 | |||
BC062591.1 | AAH62591.1 | |||
DA957269.1 | ||||
M14766.1 | AAA52435.1 | |||
M15059.1 | AAA52434.1 | |||
X04772.1 | CAA28465.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833731 | Chr.19:7589114 - 7739331 on Build GRCh38 | Loss | STXBP2 CLEC4G PET100 PCP2 FCER2 MIR6792 XAB2 TRAPPC5 MCEMP1 CAMSAP3 RETN |
nsv1060887 | Chr.19:7089546 - 7861087 on Build GRCh38 | Gain | LOC107985283 INSR CLEC4G PET100 ZNF358 PCP2 XAB2 TRAPPC5 C19orf45 MCEMP1 CLEC4M CAMSAP3 CD209 ARHGEF18 PNPLA6 STXBP2 FCER2 MIR6792 LOC100128573 MCOLN1 PEX11G CLEC4GP1 EVI5L RETN |
nsv833732 | Chr.19:7672656 - 7871329 on Build GRCh38 | Gain | PRR36 CLEC4G FCER2 TRAPPC5 MCEMP1 CLEC4M CD209 CLEC4GP1 EVI5L |
esv2751809 | Chr.19:6896369 - 7816144 on Build GRCh38 | Gain | ADGRE4P INSR CLEC4G PET100 ZNF358 MBD3L5 C19orf45 MBD3L3 CAMSAP3 PNPLA6 STXBP2 FCER2 LOC100128573 ADGRE1 LOC107985283 PCP2 XAB2 TRAPPC5 FLJ25758 MCEMP1 CLEC4M CD209 MBD3L2 ARHGEF18 ZNF557 MIR6792 LOC729458 MBD3L4 MCOLN1 PEX11G CLEC4GP1 RETN |
nsv1061452 | Chr.19:7657750 - 7785131 on Build GRCh38 | Gain | CLEC4G FCER2 TRAPPC5 MCEMP1 CLEC4M CD209 RETN |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |