Assay Details
Target Gene Details
Entrez Gene ID: | 767 |
Gene Name: | carbonic anhydrase 8 |
Gene Aliases: |
CA-RP, CA-VIII, CALS, CAMRQ3, CARP |
Location: |
Chr.8:60185420-60281423 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CA8 | NM_001321839.1 | 8 | 1203 | NP_001308768.1 |
NM_004056.5 | 9 | 1299 | NP_004047.3 | |
NR_135821.1 | 10 | 1349 | ||
XM_017013818.1 | 10 | 1364 | XP_016869307.1 | |
AK090655.1 | 9 | 1271 | ||
AK293999.1 | 8 | 890 | ||
AY075022.1 | 9 | 1276 | AAL78170.1 | |
BC069744.1 | 9 | 1268 | AAH69744.1 | |
BC069794.1 | 9 | 1303 | AAH69794.1 | |
BC108929.2 | 9 | 1224 | AAI08930.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831332 | Chr.8:60073041 - 60245171 on Build GRCh38 | Gain | CA8 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |