Assay Details
Target Gene Details
Entrez Gene ID: | 56474 |
Gene Name: | CTP synthase 2 |
Gene Aliases: |
- |
Location: |
Chr.X:16587999-16712979 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CTPS2 | NM_001144002.1 | 19 | 2942 | NP_001137474.1 |
NM_019857.4 | 19 | 2495 | NP_062831.3 | |
NM_175859.2 | 19 | 2581 | NP_787055.1 | |
XM_005274562.3 | 19 | 2358 | XP_005274619.1 | |
XM_006724503.3 | 20 | 2533 | XP_006724566.1 | |
XM_011545545.2 | 19 | 2354 | XP_011543847.1 | |
AK024070.1 | 19 | 2436 | BAB14814.1 | |
AK025654.1 | 7 | 947 | BAB15204.1 | |
AK125332.1 | 19 | 2942 | ||
AK309508.1 | 4 | 998 | ||
BC034986.1 | 19 | 2560 | AAH34986.1 | |
BX647712.1 | 2 | 938 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv438125 | Chr.X:16585228 - 16592399 on Build GRCh38 | Loss | CTPS2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |