Assay Details
Target Gene Details
Entrez Gene ID: | 7398 |
Gene Name: | ubiquitin specific peptidase 1 |
Gene Aliases: |
UBP |
Location: |
Chr.1:62436304-62451804 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
USP1 | NM_001017415.1 | 9 | 2905 | NP_001017415.1 |
NM_001017416.1 | 9 | 2810 | NP_001017416.1 | |
NM_003368.4 | 9 | 3335 | NP_003359.3 | |
AB014458.1 | 9 | 2686 | BAA34703.1 | |
AB208893.1 | 9 | 2966 | BAD92130.1 | |
AF117386.1 | 9 | 2822 | AAD11441.1 | |
AK096788.1 | 1 | 1617 | ||
AL117503.1 | 1 | 874 | CAB55967.1 | |
AL117575.1 | 8 | 2657 | CAB55999.1 | |
BC050525.1 | 9 | 3039 | AAH50525.1 | |
BQ186837.1 | 1 | 82 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |