Assay Details
Target Gene Details
Entrez Gene ID: | 151258 |
Gene Name: | solute carrier family 38 member 11 |
Gene Aliases: |
AVT2 |
Location: |
Chr.2:164897745-164955721 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC38A11 | NM_001199148.1 | 10 | 1207 | NP_001186077.1 |
NM_173512.2 | 9 | 1141 | NP_775783.1 | |
XM_005246350.4 | 11 | 1360 | XP_005246407.1 | |
XM_006712337.3 | 10 | 1356 | XP_006712400.1 | |
XM_011510737.2 | 10 | 1452 | XP_011509039.1 | |
XM_011510742.2 | 8 | 1148 | XP_011509044.1 | |
XM_017003455.1 | 11 | 1596 | XP_016858944.1 | |
XM_017003456.1 | 10 | 1530 | XP_016858945.1 | |
XM_017003457.1 | 10 | 1521 | XP_016858946.1 | |
XM_017003458.1 | 10 | 1481 | XP_016858947.1 | |
XM_017003459.1 | 9 | 1219 | XP_016858948.1 | |
XM_017003460.1 | 11 | 1596 | XP_016858949.1 | |
XM_017003461.1 | 11 | 1596 | XP_016858950.1 | |
XM_017003462.1 | 11 | 1596 | XP_016858951.1 | |
XM_017003463.1 | 10 | 993 | XP_016858952.1 | |
XM_017003464.1 | 7 | 1205 | XP_016858953.1 | |
XM_017003465.1 | 6 | 733 | XP_016858954.1 | |
AK097141.1 | 9 | 1141 | BAC04962.1 | |
AK293998.1 | 10 | 1168 | ||
BC125157.1 | 12 | 1134 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834444 | Chr.2:164865781 - 165038205 on Build GRCh38 | Gain | SLC38A11 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |