Assay Details
Target Gene Details
Entrez Gene ID: | 9622 |
Gene Name: | kallikrein related peptidase 4 |
Gene Aliases: |
AI2A1, ARM1, EMSP, EMSP1, KLK-L1, PRSS17, PSTS, kallikrein |
Location: |
Chr.19:50906352-50910745 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLK4 | NM_001302961.1 | NP_001289890.1 | ||
NM_004917.4 | NP_004908.4 | |||
NR_126566.1 | ||||
XM_011527545.2 | XP_011525847.1 | |||
XM_011527546.2 | XP_011525848.1 | |||
AF113140.1 | AAD21580.1 | |||
AF126401.1 | AAG43246.1 | |||
AF259964.1 | AAK71700.1 | |||
AF259965.1 | AAK71701.1 | |||
AF259966.1 | AAK71702.1 | |||
AF259968.1 | AAK71704.1 | |||
AF259969.1 | AAF81227.1 | |||
AF259970.1 | AAK71705.1 | |||
AF259971.1 | AAK71706.1 | |||
AF464755.1 | AAQ04828.1 | |||
AK309958.1 | ||||
AY923170.1 | AAX30051.1 | |||
BC069325.1 | AAH69325.1 | |||
BC069403.1 | AAH69403.1 | |||
BC069429.1 | AAH69429.1 | |||
BC069489.1 | AAH69489.1 | |||
BC096175.1 | AAH96175.1 | |||
BC096176.3 | AAH96176.1 | |||
BC096177.3 | ||||
BC096178.3 | AAH96178.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3583473 | Chr.19:50906008 - 50922148 on Build GRCh38 | Loss | KLK4 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |