Assay Details
Target Gene Details
Entrez Gene ID: | 57611 |
Gene Name: | immunoglobulin superfamily containing leucine rich repeat 2 |
Gene Aliases: |
LINX |
Location: |
Chr.15:74128203-74141826 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ISLR2 | NM_001130136.1 | 4 | 1565 | NP_001123608.1 |
NM_001130137.1 | 4 | 1301 | NP_001123609.1 | |
NM_001130138.1 | 4 | 1009 | NP_001123610.1 | |
NM_020851.2 | 3 | 1134 | NP_065902.1 | |
XM_011521840.2 | 4 | 1404 | XP_011520142.1 | |
XM_011521841.1 | 4 | 1087 | XP_011520143.1 | |
XM_017022446.1 | 3 | 1236 | XP_016877935.1 | |
AB040898.2 | 4 | 1565 | ||
AK092297.1 | 3 | 960 | ||
AK092302.1 | 4 | 1581 | ||
AK096602.1 | 2 | 912 | ||
AK290467.1 | 3 | 1003 | ||
AY358316.1 | 3 | 932 | AAQ88682.1 | |
BC114551.1 | AAI14552.1 | |||
BC152429.1 | 4 | 1565 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833053 | Chr.15:74067815 - 74262799 on Build GRCh38 | Loss | GOLGA6A CCDC33 LOC283731 LOC732265 STRA6 ISLR ISLR2 |
nsv1050884 | Chr.15:74102642 - 74197072 on Build GRCh38 | Gain | LOC283731 STRA6 ISLR ISLR2 |
nsv469619 | Chr.15:73999518 - 74173980 on Build GRCh38 | Loss | GOLGA6A PML LOC283731 LOC732265 ISLR ISLR2 |
nsv827385 | Chr.15:74127198 - 74136960 on Build GRCh38 | Gain | LOC283731 ISLR2 |
nsv951993 | Chr.15:74124460 - 74137359 on Build GRCh38 | Deletion | LOC283731 ISLR2 |
esv2760040 | Chr.15:73922773 - 74172575 on Build GRCh38 | Gain+Loss | GOLGA6A STOML1 LOXL1-AS1 PML LOC283731 LOC732265 ISLR2 LOXL1 |
nsv1047564 | Chr.15:74116495 - 74165840 on Build GRCh38 | Gain | LOC283731 ISLR2 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | PPCDC LOXL1-AS1 CYP1A1 LOC102723750 CLK3 CYP1A2 RPP25 COX5A UBL7-AS1 MIR6882 LMAN1L MIR4513 MPI C15orf59-AS1 MIR6881 HIGD2B REC114 UBL7 LOC105376731 TBC1D21 EDC3 ARID3B ADPGK PML LOC283731 SCAMP2 LOXL1 GOLGA6A BBS4 NPTN NEO1 LOC107984728 NPTN-IT1 CD276 STRA6 HCN4 CPLX3 GOLGA6C CCDC33 ADPGK-AS1 SEMA7A LOC732265 ISLR LOC729739 ISLR2 CYP11A1 ULK3 FAM219B C15orf39 CSK SCAMP5 LOC101929333 C15orf59 STOML1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |