Assay Details
Target Gene Details
Entrez Gene ID: | 37 |
Gene Name: | acyl-CoA dehydrogenase, very long chain |
Gene Aliases: |
ACAD6, LCACD, VLCAD |
Location: |
Chr.17:7217125-7225267 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACADVL | NM_001270447.1 | NP_001257376.1 | ||
AK293549.1 |
Target Gene Details
Entrez Gene ID: | 1742 |
Gene Name: | discs large MAGUK scaffold protein 4 |
Gene Aliases: |
PSD95, SAP-90, SAP90 |
Location: |
Chr.17:7189890-7220050 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DLG4 | NM_001321074.1 | 1 | 976 | NP_001308003.1 |
NM_001365.4 | 1 | 976 | NP_001356.1 | |
NR_135527.1 | 1 | 976 | ||
XM_011523698.1 | 1 | 774 | XP_011522000.1 | |
XM_011523699.2 | XP_011522001.1 | |||
AK296357.1 | 1 | 664 | ||
BC136553.1 | 1 | 509 | ||
U83192.1 | 1 | 636 | AAC52113.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1962 | Chr.17:7201180 - 7234067 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() |
esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3639861 | Chr.17:7198042 - 7261345 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv952118 | Chr.17:7174582 - 7456981 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2665200 | Chr.17:7091816 - 7333062 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv574322 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)