Assay Details
Target Gene Details
Entrez Gene ID: | 323 |
Gene Name: | amyloid beta precursor protein binding family B member 2 |
Gene Aliases: |
FE65L, FE65L1 |
Location: |
Chr.4:40810027-41216714 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv10486 | Chr.4:40747595 - 40935088 on Build GRCh38 | Gain | NSUN7 APBB2 |
nsv4307 | Chr.4:40796057 - 40823222 on Build GRCh38 | Deletion | NSUN7 APBB2 |
esv3348981 | Chr.4:40814778 - 40816576 on Build GRCh38 | Duplication | APBB2 |
nsv829912 | Chr.4:40720092 - 40840600 on Build GRCh38 | Loss | NSUN7 APBB2 |
nsv999749 | Chr.4:40768481 - 40820050 on Build GRCh38 | Gain | NSUN7 APBB2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |