Assay Details
Target Gene Details
Entrez Gene ID: | 641372 |
Gene Name: | acyl-CoA thioesterase 6 |
Gene Aliases: |
C14orf42, c14_5530 |
Location: |
Chr.14:73612276-73619929 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACOT6 | NM_001037162.1 | 2 | 891 | NP_001032239.1 |
XM_011537076.2 | 3 | 1498 | XP_011535378.1 | |
BC126378.1 | 2 | 658 | ||
BC126380.1 | 2 | 658 | ||
DQ082756.1 | 2 | 891 | AAZ31238.1 |
Target Gene Details
Entrez Gene ID: | 399671 |
Gene Name: | HEAT repeat containing 4 |
Gene Aliases: |
- |
Location: |
Chr.14:73478484-73634418 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HEATR4 | XM_017021289.1 | XP_016876778.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1053281 | Chr.14:73518500 - 74050155 on Build GRCh38 | Gain | MIR4505 FAM161B LOC100506476 ACOT1 ZNF410 COQ6 LOC100506498 ELMSAN1 ACOT4 ACOT2 HEATR4 BBOF1 DNAL1 PTGR2 ENTPD5 ACOT6 PNMA1 |
esv3892561 | Chr.14:73365865 - 73620531 on Build GRCh38 | Gain | C14orf169 ACOT1 HEATR4 LOC105370561 NUMB LOC101928143 ACOT6 ACOT4 ACOT2 |
esv2759996 | Chr.14:73493593 - 73660367 on Build GRCh38 | Gain | ACOT1 HEATR4 DNAL1 ACOT6 ACOT4 ACOT2 |
nsv1035460 | Chr.14:72854572 - 73897173 on Build GRCh38 | Gain | RBM25 MIR4505 ZFYVE1 DCAF4 LOC100506476 ACOT1 ZNF410 PAPLN LOC100506498 ELMSAN1 ACOT4 ACOT2 C14orf169 DPF3 HEATR4 PSEN1 LOC105370561 LOC101928123 DNAL1 PTGR2 NUMB LOC101928143 ACOT6 PNMA1 |
nsv832828 | Chr.14:73498250 - 73664448 on Build GRCh38 | Gain+Loss | ACOT1 HEATR4 DNAL1 ACOT6 ACOT4 ACOT2 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |