Assay Details
Target Gene Details
Entrez Gene ID: | 346606 |
Gene Name: | monoacylglycerol O-acyltransferase 3 |
Gene Aliases: |
DC7, DGAT2L2, MGAT3 |
Location: |
Chr.7:101192886-101201131 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MOGAT3 | NM_001287147.1 | 6 | 913 | NP_001274076.1 |
NM_178176.3 | 7 | 1116 | NP_835470.1 | |
XM_005250309.3 | 6 | 1312 | XP_005250366.1 | |
XM_011516126.2 | 6 | 1118 | XP_011514428.1 | |
XM_011516127.2 | 6 | 905 | XP_011514429.1 | |
AY229854.1 | 7 | 1116 | AAO63579.1 | |
AY358200.1 | 7 | 1093 | AAQ88567.1 | |
BC100953.1 | 7 | 1007 | AAI00954.1 | |
BC100954.1 | 6 | 1093 | AAI00955.1 | |
BC100955.1 | 6 | 807 | AAI00956.1 | |
BE042458.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422376 | Chr.7:101100204 - 101373190 on Build GRCh38 | Duplication | MIR4653 AP1S1 PLOD3 COL26A1 FIS1 CLDN15 IFT22 ZNHIT1 SERPINE1 VGF LOC101927721 NAT16 LOC101927746 MOGAT3 |
esv2422326 | Chr.7:101190168 - 101356850 on Build GRCh38 | Duplication | IFT22 ZNHIT1 PLOD3 LOC101927721 FIS1 LOC101927746 MOGAT3 CLDN15 |
nsv1030604 | Chr.7:101074451 - 101580185 on Build GRCh38 | Gain | MIR4653 TRIM56 AP1S1 PLOD3 COL26A1 FIS1 CLDN15 LINC01007 IFT22 ZNHIT1 SERPINE1 VGF LOC101927721 NAT16 LOC101927746 MOGAT3 |
dgv11534n54 | Chr.7:101160859 - 101238359 on Build GRCh38 | Loss | ZNHIT1 AP1S1 PLOD3 VGF NAT16 MOGAT3 CLDN15 |
esv2752134 | Chr.7:101156850 - 101498586 on Build GRCh38 | Gain | MIR4653 AP1S1 PLOD3 COL26A1 FIS1 CLDN15 IFT22 ZNHIT1 VGF LOC101927721 NAT16 LOC101927746 MOGAT3 |
nsv607973 | Chr.7:101139179 - 101230554 on Build GRCh38 | Loss | ZNHIT1 SERPINE1 MIR4653 AP1S1 PLOD3 VGF NAT16 MOGAT3 |
esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion | STAG3L5P-PVRIG2P-PILRB MEPCE PVRIG2P PVRIG MIR4658 MUC17 C7orf43 PCOLCE-AS1 LOC105375429 MIR106B MIR6840 ZASP ZCWPW1 SLC12A9 PCOLCE GATS TRIP6 PMS2P1 SAP25 MIR93 NAT16 MOGAT3 MOSPD3 LRCH4 TFR2 SPDYE3 PPP1R35 COPS6 AZGP1P1 MIR25 LOC102724094 ZNF3 SRRT SERPINE1 MIR6875 MUC12 ZAN GIGYF1 LAMTOR4 POP7 ACTL6B AZGP1 TRIM56 PLOD3 ZKSCAN1 ACHE GNB2 STAG3L5P NYAP1 AGFG2 CLDN15 EPO MBLAC1 PILRB VGF PILRA GAL3ST4 ZSCAN21 MIR4653 AP1S1 AP4M1 FIS1 MCM7 CNPY4 FBXO24 STAG3 TAF6 ZNHIT1 EPHB4 UFSP1 C7orf61 MUC3A TSC22D4 GPC2 |
nsv831075 | Chr.7:101049016 - 101273737 on Build GRCh38 | Loss | ZNHIT1 SERPINE1 MIR4653 TRIM56 AP1S1 PLOD3 VGF FIS1 MUC17 NAT16 MOGAT3 CLDN15 |
nsv607974 | Chr.7:101149529 - 101328323 on Build GRCh38 | Loss | IFT22 ZNHIT1 MIR4653 AP1S1 PLOD3 VGF LOC101927721 FIS1 NAT16 LOC101927746 MOGAT3 CLDN15 |
nsv1018841 | Chr.7:101117129 - 101225018 on Build GRCh38 | Gain | ZNHIT1 SERPINE1 MIR4653 AP1S1 PLOD3 VGF NAT16 MOGAT3 |
nsv831076 | Chr.7:101185566 - 101368393 on Build GRCh38 | Loss | IFT22 ZNHIT1 PLOD3 COL26A1 LOC101927721 FIS1 LOC101927746 MOGAT3 CLDN15 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |