Assay Details
Target Gene Details
Entrez Gene ID: | 79585 |
Gene Name: | coronin 7 |
Gene Aliases: |
0610011B16Rik, CRN7, POD1 |
Location: |
Chr.16:4354542-4416961 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CORO7 | NM_001201472.1 | 24 | 2796 | NP_001188401.1 |
NM_001201473.1 | 22 | 2595 | NP_001188402.1 | |
NM_024535.4 | 24 | 2850 | NP_078811.3 | |
AK025674.1 | 24 | 2459 | BAB15211.1 | |
AK094596.1 | 24 | 2469 | ||
AK097238.1 | 24 | 2316 | ||
AK126176.1 | 10 | 1622 | ||
AK294045.1 | 24 | 2431 | ||
AK296723.1 | 24 | 2428 | ||
AK296807.1 | 22 | 2261 | ||
BC117289.1 | 24 | 2469 | ||
BC117291.1 | 24 | 2469 |
Target Gene Details
Entrez Gene ID: | 100529144 |
Gene Name: | CORO7-PAM16 readthrough |
Gene Aliases: |
- |
Location: |
Chr.16:4340251-4416961 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CORO7-PAM16 | NM_001201479.1 | 24 | 2850 | NP_001188408.1 |
AL833954.1 | 26 | 2925 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471075 | Chr.16:4317729 - 4395326 on Build GRCh38 | Loss | PAM16 CORO7 GLIS2-AS1 VASN GLIS2 CORO7-PAM16 |
esv2659570 | Chr.16:4353172 - 4362927 on Build GRCh38 | Deletion | CORO7 CORO7-PAM16 |
esv3637696 | Chr.16:4353267 - 4362846 on Build GRCh38 | Loss | CORO7 CORO7-PAM16 |
nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain | NMRAL1 LINC01569 GLIS2-AS1 NUDT16L1 TFAP4 ANKS3 NAGPA SEC14L5 C16orf71 PAM16 PPL SMIM22 C16orf96 SEPT12 GLYR1 VASN GLIS2 NAGPA-AS1 MGRN1 CDIP1 UBN1 ALG1 SRL CORO7 LOC105371063 RBFOX1 ZNF500 C16orf89 HMOX2 MIR6769A DNAJA3 ADCY9 CORO7-PAM16 ROGDI EEF2KMT UBALD1 |
nsv571274 | Chr.16:4309420 - 4441189 on Build GRCh38 | Gain | PAM16 CORO7 GLIS2-AS1 VASN GLIS2 DNAJA3 CORO7-PAM16 |
nsv527866 | Chr.16:4206462 - 4412618 on Build GRCh38 | Loss | PAM16 SRL LINC01569 CORO7 LOC105371063 GLIS2-AS1 VASN GLIS2 TFAP4 CORO7-PAM16 |
esv3584924 | Chr.16:4330628 - 4469258 on Build GRCh38 | Gain | NMRAL1 PAM16 CORO7 VASN GLIS2 DNAJA3 CORO7-PAM16 |
nsv520212 | Chr.16:4354619 - 4390827 on Build GRCh38 | Loss | CORO7 VASN CORO7-PAM16 |
esv23732 | Chr.16:4340813 - 4476244 on Build GRCh38 | Gain | NMRAL1 PAM16 HMOX2 CORO7 VASN DNAJA3 CORO7-PAM16 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |