Assay Details
Target Gene Details
Entrez Gene ID: | 57611 |
Gene Name: | immunoglobulin superfamily containing leucine rich repeat 2 |
Gene Aliases: |
LINX |
Location: |
Chr.15:74128203-74141826 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ISLR2 | NM_001130136.1 | NP_001123608.1 | ||
NM_001130137.1 | NP_001123609.1 | |||
NM_001130138.1 | NP_001123610.1 | |||
NM_020851.2 | NP_065902.1 | |||
XM_011521840.2 | XP_011520142.1 | |||
XM_011521841.1 | XP_011520143.1 | |||
XM_017022446.1 | XP_016877935.1 | |||
AB040898.2 | ||||
AK092297.1 | ||||
AK092302.1 | ||||
AK290467.1 | ||||
AY358316.1 | AAQ88682.1 | |||
BC152429.1 | ||||
BI917487.1 | ||||
DA791954.1 | ||||
DA911394.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833053 | Chr.15:74067815 - 74262799 on Build GRCh38 | Loss | ISLR2 GOLGA6A ISLR LOC732265 LOC283731 STRA6 CCDC33 |
nsv1050884 | Chr.15:74102642 - 74197072 on Build GRCh38 | Gain | ISLR2 ISLR LOC283731 STRA6 |
nsv469619 | Chr.15:73999518 - 74173980 on Build GRCh38 | Loss | ISLR2 GOLGA6A PML ISLR LOC732265 LOC283731 |
nsv827385 | Chr.15:74127198 - 74136960 on Build GRCh38 | Gain | ISLR2 LOC283731 |
nsv951993 | Chr.15:74124460 - 74137359 on Build GRCh38 | Deletion | ISLR2 LOC283731 |
esv2760040 | Chr.15:73922773 - 74172575 on Build GRCh38 | Gain+Loss | ISLR2 GOLGA6A PML LOC732265 LOXL1 LOC283731 STOML1 LOXL1-AS1 |
nsv1047564 | Chr.15:74116495 - 74165840 on Build GRCh38 | Gain | ISLR2 LOC283731 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | LOC732265 ADPGK C15orf39 LOXL1 LOC102723750 GOLGA6A CYP1A1 NPTN CLK3 ULK3 BBS4 COX5A ISLR2 MPI MIR6882 ARID3B TBC1D21 RPP25 ISLR FAM219B LOC283731 CD276 LMAN1L STOML1 CCDC33 NEO1 NPTN-IT1 CYP1A2 CYP11A1 EDC3 UBL7 PML LOC105376731 SCAMP2 LOC107984728 C15orf59-AS1 ADPGK-AS1 MIR6881 CSK SCAMP5 HCN4 LOC729739 STRA6 LOC101929333 PPCDC MIR4513 LOXL1-AS1 CPLX3 SEMA7A C15orf59 HIGD2B UBL7-AS1 GOLGA6C REC114 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |