Assay Details
Target Gene Details
Entrez Gene ID: | 7398 |
Gene Name: | ubiquitin specific peptidase 1 |
Gene Aliases: |
UBP |
Location: |
Chr.1:62436304-62451804 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
USP1 | NM_001017415.1 | 2 | 332 | NP_001017415.1 |
NM_001017416.1 | 2 | 237 | NP_001017416.1 | |
NM_003368.4 | 2 | 762 | NP_003359.3 | |
AB014458.1 | 2 | 113 | BAA34703.1 | |
AB208893.1 | 2 | 394 | BAD92130.1 | |
AF117386.1 | 2 | 249 | AAD11441.1 | |
AK312423.1 | 2 | 425 | ||
AL117575.1 | 1 | 84 | CAB55999.1 | |
AL599240.1 | 2 | 244 | ||
BC018745.1 | 2 | 762 | ||
BC032364.1 | 2 | 412 | ||
BC050525.1 | 2 | 466 | AAH50525.1 | |
BI463072.1 | 2 | 341 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |