Assay Details
Target Gene Details
Entrez Gene ID: | 2946 |
Gene Name: | glutathione S-transferase mu 2 (muscle) |
Gene Aliases: |
GST4, GSTM, GSTM2-2, GTHMUS |
Location: |
Chr.1:109668022-109683997 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GSTM2 | NM_000848.3 | 5 | 410 | NP_000839.1 |
NM_001142368.1 | 5 | 410 | NP_001135840.1 | |
AK299482.1 | 5 | 539 | ||
AK309796.1 | 5 | 327 | ||
BC105038.1 | 5 | 341 | AAI05039.1 | |
BC105066.1 | 5 | 341 | AAI05067.1 | |
BC110380.1 | 5 | 363 | ||
BM924181.1 | 5 | 376 | ||
BT019947.1 | 5 | 316 | AAV38750.1 | |
DA528798.1 | 5 | 410 | ||
M63509.1 | 5 | 332 | AAA60963.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv258n100 | Chr.1:109658958 - 109700319 on Build GRCh38 | Gain |
![]() ![]() ![]() |
dgv254n100 | Chr.1:109631914 - 109700319 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() |
dgv15n111 | Chr.1:109664699 - 109703504 on Build GRCh38 | Deletion |
![]() ![]() ![]() |
dgv256n100 | Chr.1:109638683 - 109700319 on Build GRCh38 | Loss |
![]() ![]() ![]() |
esv3587153 | Chr.1:109660894 - 109674476 on Build GRCh38 | Loss |
![]() ![]() |
nsv946134 | Chr.1:109668513 - 109716699 on Build GRCh38 | Duplication |
![]() ![]() ![]() |
dgv257n100 | Chr.1:109650144 - 109707602 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv482081 | Chr.1:109668022 - 109683996 on Build GRCh38 | Gain |
![]() |
dgv77e212 | Chr.1:109635559 - 109709330 on Build GRCh38 | Loss |
![]() ![]() ![]() |
dgv259n100 | Chr.1:109663341 - 109700319 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() |
nsv547486 | Chr.1:109657738 - 109690459 on Build GRCh38 | Loss |
![]() ![]() ![]() |
esv25172 | Chr.1:109644224 - 109716486 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() |
dgv253n100 | Chr.1:109613590 - 109700319 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)