Assay Details
Target Gene Details
Entrez Gene ID: | 116461 |
Gene Name: | tRNA splicing endonuclease subunit 15 |
Gene Aliases: |
C1orf19, PCH2F, sen15 |
Location: |
Chr.1:184050207-184095846 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TSEN15 | NM_001127394.3 | 4 | 1128 | NP_001120866.1 |
NM_001300764.1 | 5 | 1282 | NP_001287693.1 | |
NM_001300766.1 | 5 | 1166 | NP_001287695.1 | |
NM_052965.3 | 5 | 1270 | NP_443197.1 | |
NR_023349.2 | 6 | 1387 | ||
NR_125335.1 | 6 | 1350 | ||
XM_011509139.2 | XP_011507441.1 | |||
XM_017000228.1 | XP_016855717.1 | |||
XM_017000229.1 | XP_016855718.1 | |||
XM_017000230.1 | XP_016855719.1 | |||
AF288394.1 | 5 | 1245 | AAG60614.1 | |
AK098196.1 | 5 | 1168 | ||
AK296655.1 | 4 | 1023 | ||
AK307241.1 | 6 | 1278 | ||
BC022030.1 | 5 | 1215 | AAH22030.1 | |
BM789892.1 | ||||
CB158796.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |