Assay Details
Target Gene Details
Entrez Gene ID: | 64689 |
Gene Name: | golgi reassembly stacking protein 1 |
Gene Aliases: |
GOLPH5, GRASP65, P65 |
Location: |
Chr.3:39096599-39108363 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GORASP1 | NM_001278789.1 | 5 | 911 | NP_001265718.1 |
NM_001278790.1 | 4 | 731 | NP_001265719.1 | |
NM_031899.3 | 7 | 1659 | NP_114105.1 | |
NR_103866.1 | 7 | 1155 | ||
NR_103867.1 | 7 | 1147 | ||
XM_006713301.3 | 7 | 1759 | XP_006713364.1 | |
XM_011534020.2 | 7 | 1669 | XP_011532322.1 | |
XM_011534021.2 | 6 | 1579 | XP_011532323.1 | |
XM_017007050.1 | 7 | 845 | XP_016862539.1 | |
XM_017007051.1 | 6 | 1194 | XP_016862540.1 | |
XM_017007052.1 | 6 | 790 | XP_016862541.1 | |
AJ409349.1 | 7 | 935 | CAC35160.1 | |
AK001574.1 | 6 | 1177 | ||
AK057043.1 | 4 | 469 | ||
AK091168.1 | 7 | 1123 | BAC03598.1 | |
AK124755.1 | 7 | 1659 | ||
AK300271.1 | ||||
AK303847.1 | 5 | 651 | ||
BC008928.2 | 7 | 876 | AAH08928.1 | |
BC075854.1 | 7 | 945 | AAH75854.1 | |
BC103826.1 | 7 | 877 | AAI03827.1 | |
BC103827.1 | 7 | 877 | AAI03828.1 | |
BM470731.1 | 2 | 172 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |