Assay Details
Target Gene Details
Entrez Gene ID: | 63908 |
Gene Name: | NSF attachment protein beta |
Gene Aliases: |
SNAP-BETA, SNAPB |
Location: |
Chr.20:23374519-23421519 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NAPB | NM_001283018.1 | 11 | 2966 | NP_001269947.1 |
NM_001283020.1 | 10 | 2837 | NP_001269949.1 | |
NM_001283026.1 | 10 | 2907 | NP_001269955.1 | |
NM_022080.2 | 11 | 2954 | NP_071363.1 | |
NR_104266.1 | 11 | 2481 | ||
XM_011529313.1 | 11 | 2958 | XP_011527615.1 | |
XM_017028008.1 | 11 | 3161 | XP_016883497.1 | |
AK022817.1 | 7 | 2480 | ||
AK124876.1 | 11 | 2893 | ||
AK125811.1 | 4 | 3813 | ||
AK126340.1 | 7 | 4304 | ||
AL834445.1 | 5 | 2370 | CAD39105.1 | |
BC047426.1 | 10 | 2849 | AAH47426.1 | |
BC060840.1 | 11 | 2879 | AAH60840.1 | |
BX648330.1 | 11 | 2437 |
More Information
Additional Information:
For this assay, SNP(s) [rs74892021] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Exonic non-DGV Variation |