Assay Details
Target Gene Details
Entrez Gene ID: | 64223 |
Gene Name: | MTOR associated protein, LST8 homolog |
Gene Aliases: |
GBL, GbetaL, LST8, POP3, WAT1 |
Location: |
Chr.16:2205177-2209417 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MLST8 | NM_001199173.1 | NP_001186102.1 | ||
NM_001199174.1 | NP_001186103.1 | |||
NM_001199175.1 | NP_001186104.1 | |||
NM_022372.4 | NP_071767.3 | |||
XM_005255475.3 | XP_005255532.2 | |||
XM_005255478.2 | XP_005255535.1 | |||
XM_005255479.2 | XP_005255536.1 | |||
XM_017023548.1 | XP_016879037.1 | |||
XM_017023549.1 | XP_016879038.1 | |||
XM_017023550.1 | XP_016879039.1 | |||
XM_017023551.1 | XP_016879040.1 | |||
XM_017023552.1 | XP_016879041.1 | |||
AK021536.1 | ||||
AK022227.1 | BAB13990.1 | |||
AK098762.1 | ||||
AK302201.1 | ||||
AK304390.1 | ||||
AU117583.1 | ||||
AU121635.1 | ||||
AY223837.1 | AAO73410.1 | |||
BC001313.2 | AAH01313.1 | |||
BC017119.1 | AAH17119.1 | |||
BC052292.1 | AAH52292.1 | |||
BC088354.1 | AAH88354.1 | |||
BI761765.1 | ||||
DA382215.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | CASKIN1 LOC107984876 ERVK13-1 AMDHD2 TRAF7 SSTR5 MRPS34 FAHD1 PDPK1 FAM173A POLR3K HBA1 PTX4 MIR6859-4 FBXL16 BRICD5 LOC105371038 MIR3180-5 MIR6768 WASIR2 NPW NDUFB10 MSRB1 MAPK8IP3 C1QTNF8 CCDC154 CLCN7 ZNF598 PRSS29P IGFALS RNF151 LOC105371184 MIR4717 WFIKKN1 SNORA64 NPRL3 JMJD8 RAB26 MIR940 SPSB3 LOC107987233 LINC00235 NME3 HN1L HAGHL RNPS1 TBL3 HBM LOC106660606 NUBP2 MRPL28 CAPN15 E4F1 RHBDL1 PRR35 WDR24 MIR3178 TSR3 HBQ1 IFT140 SNHG19 TPSB2 NME4 METTL26 TPSG1 ARHGDIG RPL23AP5 HBA2 SNORD60 LOC100134368 LOC729652 LOC105371049 GFER MLST8 TSC2 KCTD5 SOX8 MEIOB SNORA78 RGS11 NTHL1 PIGQ ABCA17P EME2 CACNA1H SNRNP25 MIR3677 TMEM8A HBZ C16orf59 LOC100287175 LINC00254 LMF1-AS1 MIR3177 NHLRC4 FLJ42627 TPSD1 METRN AXIN1 MPG DDX11L10 ATP6V0C MCRIP2 CHTF18 RAB40C RPUSD1 HS3ST6 HAGH FAM234A RPS2 PRR25 LUC7L CCNF LOC101929280 NOXO1 CRAMP1 PGP CEMP1 MIR6767 RAB11FIP3 UBE2I NARFL MIR662 MIR6511B1 SNORA10 SNHG9 LOC105371046 MIR3176 C16orf91 LOC652276 RHOT2 GNG13 UNKL MIR1225 STUB1 LOC105371045 WDR90 ABCA3 RHBDF1 MIR5587 GNPTG TBC1D24 LMF1 LOC105371047 ECI1 RPL3L NTN3 SLC9A3R2 PKD1 PRSS27 TMEM204 MSLN DNASE1L2 PDIA2 SYNGR3 MIR4516 DECR2 TELO2 SSTR5-AS1 BAIAP3 CCDC78 TPSAB1 |
nsv471072 | Chr.16:1971892 - 2317508 on Build GRCh38 | Loss | MIR6511B1 CASKIN1 MIR3677 RAB26 MIR940 TRAF7 RNPS1 TBL3 MIR1225 LOC106660606 ABCA3 E4F1 BRICD5 MIR3180-5 NPW ECI1 SNHG19 SLC9A3R2 PKD1 DNASE1L2 SNORD60 SYNGR3 MIR4516 NOXO1 PGP LOC105371049 ZNF598 GFER MLST8 TSC2 MIR4717 NTHL1 |
dgv4861n54 | Chr.16:2083900 - 2246850 on Build GRCh38 | Loss | ECI1 MIR6511B1 CASKIN1 RAB26 SNHG19 TRAF7 PKD1 DNASE1L2 SNORD60 MIR4516 PGP MIR1225 LOC105371049 E4F1 MLST8 BRICD5 TSC2 MIR3180-5 |
dgv4863n54 | Chr.16:2141733 - 2274302 on Build GRCh38 | Loss | ECI1 CASKIN1 MIR3677 RAB26 SNHG19 MIR940 TRAF7 DNASE1L2 SNORD60 RNPS1 PGP LOC106660606 E4F1 MLST8 BRICD5 |
nsv1711 | Chr.16:2199093 - 2232650 on Build GRCh38 | Insertion | PGP E4F1 MLST8 BRICD5 |
nsv457351 | Chr.16:2156590 - 2274302 on Build GRCh38 | Loss | PGP ECI1 CASKIN1 LOC106660606 MIR3677 MIR940 TRAF7 E4F1 MLST8 BRICD5 DNASE1L2 RNPS1 |
nsv1052260 | Chr.16:2074546 - 2629655 on Build GRCh38 | Gain | MIR6511B1 ABCA17P CASKIN1 MIR3677 RAB26 MIR940 AMDHD2 TRAF7 C16orf59 LOC652276 RNPS1 PDPK1 MIR1225 LOC106660606 ABCA3 E4F1 BRICD5 MIR3180-5 MIR6768 TBC1D24 MIR3178 ATP6V0C ECI1 NTN3 SNHG19 PKD1 DNASE1L2 SNORD60 CCNF MIR4516 PGP LOC729652 CEMP1 MIR6767 LOC105371049 MLST8 TSC2 MIR4717 |
nsv952902 | Chr.16:2143400 - 2339999 on Build GRCh38 | Deletion | ECI1 CASKIN1 MIR3677 RAB26 SNHG19 MIR940 TRAF7 DNASE1L2 SNORD60 RNPS1 PGP LOC106660606 ABCA3 E4F1 MLST8 BRICD5 MIR4717 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |