Assay Details
Target Gene Details
Entrez Gene ID: | 5320 |
Gene Name: | phospholipase A2 group IIA |
Gene Aliases: |
MOM1, PLA2, PLA2B, PLA2L, PLA2S, PLAS1, sPLA2 |
Location: |
Chr.1:19975431-19980439 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLA2G2A | NM_000300.3 | NP_000291.1 | ||
NM_001161727.1 | NP_001155199.1 | |||
NM_001161728.1 | NP_001155200.1 | |||
NM_001161729.1 | NP_001155201.1 | |||
AK291302.1 | ||||
AY656695.1 | AAT73043.1 | |||
AY656696.1 | AAT73044.1 | |||
BC005919.1 | AAH05919.1 | |||
BM708748.1 | ||||
BU626291.1 | ||||
BX641714.1 | ||||
CB132375.1 | ||||
CR456865.1 | CAG33146.1 | |||
M22430.1 | AAA36550.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |