Assay Details
Target Gene Details
Entrez Gene ID: | 2954 |
Gene Name: | glutathione S-transferase zeta 1 |
Gene Aliases: |
GSTZ1-1, MAAI, MAI |
Location: |
Chr.14:77320887-77331597 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GSTZ1 | NM_001312660.1 | NP_001299589.1 | ||
NM_145870.2 | NP_665877.1 | |||
NM_145871.2 | NP_665878.2 | |||
XM_011536670.2 | XP_011534972.1 | |||
AB209360.1 | BAD92597.1 | |||
AJ001838.1 | CAA05045.1 | |||
AK309054.1 | ||||
AK315154.1 | ||||
AK315609.1 | ||||
BC001453.1 | AAH01453.1 | |||
BG330365.1 | ||||
BG772450.1 | ||||
BI559916.1 | ||||
BI829992.1 | ||||
U86529.1 | AAB96392.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |