Assay Details
Target Gene Details
Entrez Gene ID: | 100131390 |
Gene Name: | Sp9 transcription factor |
Gene Aliases: |
ZNF990 |
Location: |
Chr.2:174335093-174337540 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SP9 | NM_001145250.1 | NP_001138722.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv583666 | Chr.2:174324901 - 174346560 on Build GRCh38 | Loss | LINC01305 SP9 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |