Assay Details
Target Gene Details
Entrez Gene ID: | 55788 |
Gene Name: | LMBR1 domain containing 1 |
Gene Aliases: |
C6orf209, LMBD1, MAHCF, NESI |
Location: |
Chr.6:69675749-69797157 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LMBRD1 | NM_018368.3 | 15 | 1748 | NP_060838.3 |
XM_011535941.1 | 15 | 1598 | XP_011534243.1 | |
AF113224.1 | 15 | 1468 | AAG39295.1 | |
AF208863.1 | 14 | 1596 | AAF64277.1 | |
AF211480.1 | 12 | 1086 | AAK26247.1 | |
AK002102.1 | 15 | 1497 | BAA92087.1 | |
AK225187.1 | 15 | 1497 | ||
AK225537.1 | 15 | 1482 | ||
AK290069.1 | 15 | 1628 | ||
AY136817.1 | 14 | 1253 | AAN11301.1 | |
BC010360.1 | 13 | 1247 | AAH10360.1 | |
BC047073.1 | 15 | 1587 | AAH47073.1 | |
BX647737.1 | 14 | 1340 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |