Assay Details
Target Gene Details
Entrez Gene ID: | 11234 |
Gene Name: | HPS5, biogenesis of lysosomal organelles complex 2 subunit 2 |
Gene Aliases: |
AIBP63, BLOC2S2 |
Location: |
Chr.11:18278670-18322498 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HPS5 | NM_007216.3 | 22 | 3656 | NP_009147.3 |
NM_181507.1 | 23 | 3813 | NP_852608.1 | |
NM_181508.1 | 22 | 3469 | NP_852609.1 | |
XM_011519862.1 | 24 | 3938 | XP_011518164.1 | |
XM_011519863.1 | 24 | 3846 | XP_011518165.1 | |
XM_011519864.1 | 24 | 3778 | XP_011518166.1 | |
XM_011519865.1 | 23 | 4160 | XP_011518167.1 | |
XM_011519866.1 | 23 | 3811 | XP_011518168.1 | |
XM_011519867.1 | 23 | 3677 | XP_011518169.1 | |
XM_011519868.2 | 23 | 3918 | XP_011518170.1 | |
XM_017017149.1 | 23 | 3712 | XP_016872638.1 | |
XM_017017150.1 | 23 | 3644 | XP_016872639.1 | |
XM_017017151.1 | 23 | 3996 | XP_016872640.1 | |
XM_017017152.1 | 22 | 4026 | XP_016872641.1 | |
XM_017017153.1 | 22 | 3862 | XP_016872642.1 | |
XM_017017154.1 | 22 | 3543 | XP_016872643.1 | |
AB023234.1 | 22 | 3469 | ||
AF534400.1 | 22 | 3469 | AAO25962.1 | |
AF534401.1 | 23 | 3813 | AAO25963.1 | |
AF534402.1 | 22 | 3656 | AAO25964.1 | |
AJ131721.1 | 11 | 1987 | CAB38232.1 | |
AK024320.1 | 8 | 1596 | ||
AK292436.1 | ||||
BC033640.1 | 22 | 3617 | AAH33640.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |