Assay Details
Target Gene Details
Entrez Gene ID: | 199223 |
Gene Name: | tetratricopeptide repeat domain 21A |
Gene Aliases: |
IFT139A, STI2 |
Location: |
Chr.3:39107661-39139009 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TTC21A | NM_001105513.2 | 14 | 2073 | NP_001098983.2 |
NM_145755.2 | 15 | 2217 | NP_665698.2 | |
XM_005264921.4 | 15 | 2146 | XP_005264978.1 | |
XM_005264922.4 | 15 | 2146 | XP_005264979.1 | |
XM_005264923.3 | 15 | 2146 | XP_005264980.1 | |
XM_005264924.4 | 15 | 2146 | XP_005264981.1 | |
XM_005264925.4 | 14 | 2023 | XP_005264982.1 | |
XM_005264926.4 | 15 | 2146 | XP_005264983.1 | |
XM_005264927.4 | 11 | 1600 | XP_005264984.1 | |
XM_006713011.3 | 15 | 2146 | XP_006713074.1 | |
XM_011533447.2 | 14 | 2047 | XP_011531749.1 | |
XM_011533449.1 | 15 | 2146 | XP_011531751.1 | |
XM_017005843.1 | 15 | 2146 | XP_016861332.1 | |
AJ487015.1 | 15 | 2179 | CAD31647.1 | |
AK093313.1 | 5 | 757 | BAC04129.1 | |
AK302408.1 | 14 | 2056 | ||
AK302465.1 | 14 | 1927 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |