Assay Details
Target Gene Details
Entrez Gene ID: | 2825 |
Gene Name: | G protein-coupled receptor 1 |
Gene Aliases: |
- |
Location: |
Chr.2:206175316-206218047 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPR1 | NM_001098199.1 | 3 | 1141 | NP_001091669.1 |
NM_001261452.1 | 4 | 1392 | NP_001248381.1 | |
NM_001261453.1 | 4 | 1396 | NP_001248382.1 | |
NM_001261454.1 | 3 | 987 | NP_001248383.1 | |
NM_001261455.1 | 3 | 983 | NP_001248384.1 | |
NM_005279.3 | 3 | 1145 | NP_005270.2 | |
XM_005246471.2 | 3 | 907 | XP_005246528.1 | |
XM_017003832.1 | 3 | 1844 | XP_016859321.1 | |
AK075130.1 | 2 | 852 | ||
AK290976.1 | 3 | 1059 | ||
AK312917.1 | 4 | 1248 | ||
AL550780.3 | 2 | 840 | ||
BC067833.1 | 3 | 1010 | AAH67833.1 | |
BX094581.1 | 1 | 378 | ||
BX389743.2 | ||||
EF577403.1 | 1 | 759 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |