Assay Details
Target Gene Details
Entrez Gene ID: | 4793 |
Gene Name: | NFKB inhibitor beta |
Gene Aliases: |
IKBB, TRIP9 |
Location: |
Chr.19:38899503-38908894 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NFKBIB | NM_001243116.1 | NP_001230045.1 | ||
NM_002503.4 | NP_002494.2 | |||
NR_040515.1 | 5 | 1237 | ||
XM_006723226.2 | XP_006723289.2 | |||
XM_006723227.3 | XP_006723290.3 | |||
AB209784.1 | 5 | 1259 | BAD93021.1 | |
AI684894.1 | ||||
AK057862.1 | 5 | 1188 | BAB71599.1 | |
BC007197.1 | 3 | 893 | AAH07197.1 | |
BC015528.1 | AAH15528.1 | |||
BI520169.1 | ||||
BT006743.1 | AAP35389.1 | |||
BU150265.1 | 2 | 516 | ||
BU634404.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv820174 | Chr.19:38907812 - 38908108 on Build GRCh38 | Gain | NFKBIB |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |