Assay Details
Target Gene Details
Entrez Gene ID: | 1009 |
Gene Name: | cadherin 11 |
Gene Aliases: |
CAD11, CDHOB, OB, OSF-4 |
Location: |
Chr.16:64943753-65126112 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDH11 | NM_001308392.1 | 14 | 2943 | NP_001295321.1 |
NM_001797.3 | 13 | 2764 | NP_001788.2 | |
XM_005255761.3 | 13 | 2635 | XP_005255818.1 | |
XM_005255762.2 | 13 | 2849 | XP_005255819.1 | |
XM_005255763.2 | 13 | 2559 | XP_005255820.1 | |
XM_011522803.2 | 12 | 2469 | XP_011521105.1 | |
XM_017022830.1 | 12 | 2324 | XP_016878319.1 | |
AB209760.1 | 11 | 2242 | BAD92997.1 | |
AK291251.1 | 13 | 2690 | ||
AK297377.1 | 13 | 2275 | ||
BC013609.1 | 11 | 2383 | AAH13609.1 | |
D21254.1 | 13 | 2650 | BAA04798.1 | |
D21255.1 | 14 | 2864 | BAA04799.1 | |
L34056.1 | 11 | 2364 | AAA35622.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv572901 | Chr.16:64930330 - 64956284 on Build GRCh38 | Loss | CDH11 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |